Merck
  • The fentanyl/etomidate-anaesthetised beagle (FEAB) dog: a versatile in vivo model in cardiovascular safety research.
The fentanyl/etomidate-anaesthetised beagle (FEAB) dog: a versatile in vivo model in cardiovascular safety research.
Journal of pharmacological and toxicological methods (2009-05-09)
Bruno Van Deuren, Karel Van Ammel, Yves Somers, Frank Cools, Roel Straetemans, Henk J van der Linde, David J Gallacher
ABSTRACT
The purpose of conducting cardiovascular safety pharmacology studies is to investigate the pharmacological profiles of new molecular entities (NMEs) and provide data that can be used for optimization of a possible new drug, and help make a selection of NMEs for clinical development. An anaesthetised dog preparation has been used for more than two decades by our department to measure multiple cardiovascular and respiratory parameters and to evaluate different scientific models, leading to more in-depth evaluation of drug-induced cardiovascular effects. An anaesthetic regime developed in house (induction with lofentanil, scopolamine and succinylcholine, and maintenance with fentanyl and etomidate) gives us a preparation free of pain and stress, with minimal effects on the cardiovascular system. This anaesthetic regime had minimal influences on circulating catecholamine levels, on the baroreflex sensitivity, and on all measured basal parameters compared to conscious dogs. All parameters were stable for at least 3 h, with acceptable tolerance intervals, evaluated over 99 safety studies with 3 vehicle treatments (saline, 10% and 20% hydroxypropyl-beta-cyclodextrin). This translates into a highly sensitive model for detecting possible drug-induced effects of NMEs with different mechanisms of action such as: Ca-, Na-, I(Kr)-, I(Ks)-channel blockers, K- and Ca-channel activators, alpha1- and beta-agonists, and muscarinic antagonists. Fentanyl in combination with etomidate is a successful anaesthetic regime in humans [Stockham, R.J., Stanley, T.H., Pace, N.L., King, K., Groen, F. & Gillmor, S.T. (1987). Induction of anaesthesia with fentanyl or fentanyl plus etomidate in high-risk patients. Journal of Cardiothoracic Anesthesia. 1(1), 19-23.]. In the anaesthetised dog, QT correction factors (Van de Water correction and body temperature correction) and risk factors (total, short-term and long-term instability) have been evaluated, using this regime [Van de Water, A., Verheyen, J., Xhonneux, R. & Reneman, R. (1989). An improved method to correct the QT interval of the electrocardiogram for changes in heart rate. Journal of Pharmacological Methods, 22, 207-217.; van der Linde, H.J., Van Deuren, B., Teisman, A., Towart, R. & Gallacher, D.J. (2008). The effect of changes in core body temperature on the QT interval in beagle dogs: A previously ignored phenomenon, with a method for correction. British Journal of Pharmacology, 154, 1474-1481.; van der Linde, H.J., Van de Water, A., Loots, W., Van Deuren, B., Lu, H.R., Van Ammel, K., et al. (2005) A new method to calculate the beat-to-beat instability of QT duration in drug-induced long QT in anaesthetised dogs. Journal of Pharmacological and Toxicological Methods, 52, 168-177.]. Furthermore, this anaesthetic protocol has been used to create different scientific models (long QT, short QT) with different specific end-points (ventricular fibrillation, adrenergic- or pause-dependent TdP) and also their specific precursors: e.g. aftercontractions, phase 2 EADs, phase 3 EADs, DADs, T-wave morphology changes, T-wave alternans, R-on-T, transmural and interventricular dispersion [Gallacher, D.J., Van de Water, A., van der Linde, H.J., Hermans, A.N., Lu, H.R., Towart, R., et al. (2007). In vivo mechanisms precipitating torsade de pointes in canine model of drug-induced long QT1 syndrome. Cardiovascular Research, 76-2, 247-256.]. This paper gives a brief overview of the stability, reproducibility, sensitivity and utility of a well-validated anaesthetised dog model.
Product Description
Etomidate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Etomidate, >98% (HPLC), powder