Merck
Chymotrypsin C mutations in chronic pancreatitis.
Journal of gastroenterology and hepatology (2011-06-03)
Jiayi Zhou, Miklós Sahin-Tóth
ABSTRACT
Chronic pancreatitis is a persistent inflammatory disorder characterized by destruction of the pancreatic parenchyma, maldigestion, and chronic pain. Mutations in the chymotrypsin C (CTRC) gene encoding the digestive enzyme CTRC have been shown to increase the risk of chronic pancreatitis in European and Asian populations. Here, we review the biochemical properties and physiological functions of human CTRC, summarize the functional defects associated with CTRC mutations, and discuss mechanistic models that might explain the increased disease risk in carriers.
Sigma-Aldrich
α-Chymotrypsin from bovine pancreas, suitable for protein sequencing, salt-free, lyophilized powder
Sigma-Aldrich
α-Chymotrypsin from bovine pancreas, ≥40 units/mg protein, vial of 5 mg
Sigma-Aldrich
α-Chymotrypsin from bovine pancreas, Type I-S, essentially salt-free, lyophilized powder
Sigma-Aldrich
α-Chymotrypsin from human pancreas, lyophilized powder
Sigma-Aldrich
α-Chymotrypsin from bovine pancreas, Type II, lyophilized powder, ≥40 units/mg protein
Sigma-Aldrich
α-Chymotrypsin from bovine pancreas, (TLCK treated to inactivate residual tryspin activity), Type VII, essentially salt-free, lyophilized powder, ≥40 units/mg protein
Chymotrypsin, BRP, European Pharmacopoeia (EP) Reference Standard