Merck
  • Normal prions as a new target of cobalamin (vitamin B12) in rat central nervous system.
Normal prions as a new target of cobalamin (vitamin B12) in rat central nervous system.
Clinical chemistry and laboratory medicine (2012-11-28)
Giuseppe Scalabrino, Daniela Veber
ABSTRACT
The pathogenesis of cobalamin (Cbl)-deficient (Cbl-D) neuropathy and the role of normal prions (PrPcs) in myelin maintenance are both subjects of debate. We have demonstrated that Cbl deficiency damages myelin by increasing tumor necrosis factor (TNF)-α, and decreasing epidermal growth factor (EGF) levels in the rat central nervous system (CNS). It is known that TNF-α and EGF regulate PrPc expression in vitro, and that myelin vacuolation, reactive astrocytosis and microglial activation are common to rat Cbl-D neuropathy and some prion diseases. We have shown that Cbl deficiency leads to high levels of PrPcs [particularly the octapeptide repeat (OR) domains] in the rat CNS thereby damaging the spinal cord (SC) myelin, and that chronic intra-cerebroventricular treatment with anti-OR antibodies normalizes SC myelin morphology. We have also found that PrPc levels are increased in the SC of Cbl-D rats by the time the myelin lesions appear, and that this increase is mediated by excess myelinotoxic TNF-α and prevented by EGF treatment, which has proved to be as effective as Cbl in preventing Cbl deficiency-induced lesions. Cbl stimulates PrPc mRNA-related synthesis in Cbl-D SC and duodenum, two rat tissues that are severely affected by Cbl deficiency. New PrPc synthesis is a common effect of various myelinotrophic agents, two of which (EGF and anti-TNF-α antibodies) also stimulate PrPc mRNA-related synthesis in the SC of Cbl-D rats.
Sigma-Aldrich
Vitamin B12, ≥98%
Sigma-Aldrich
Vitamin B12, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥98%
Supelco
Cyanocobalamin, pharmaceutical secondary standard, certified reference material
Cyanocobalamin, European Pharmacopoeia (EP) Reference Standard
Supelco
Cyanocobalamin (B12), analytical standard
Sigma-Aldrich
Cyanocobalamin, meets USP testing specifications
Supelco
Cyanocobalamin (Vitamin B12) solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®