Merck
  • Cadmium-based quantum dots: preparation, surface modification, and applications.
Cadmium-based quantum dots: preparation, surface modification, and applications.
Journal of nanoscience and nanotechnology (2014-04-23)
Fan Zhang, Dongmei Yi, Haizhu Sun, Hao Zhang
ABSTRACT
Cadmium-based quantum dots (QDs) have the advantage of broad UV excitation, narrow emission, bright photoluminescence (PL), and high photostability, which make them fine applications in bio-imaging, electroluminescence (EL) and photovoltaic (PV) devices, catalytic hydrogen production, sensors, etc. This review article summarizes the synthetic methods such as organometallic and aqueous approaches, and the techniques employed to prepare high-quality QDs including microwave, hydrothermal, and ultrasound. QDs and their applications are emphasized for their unique properties, especially in the preparation of near infrared (NIR). Moreover, the synthesis of novel cadmium-based QDs with metal chalcogenide complexes (MCCs) as ligands has been introduced, which can overcome traditional QDs' disadvantages of poor interparticle coupling and conducting. Following the synthetic methods, the latest development of surface modification is summarized, which is important and necessary for the applications of QDs.
Sigma-Aldrich
Cadmium, granular, 30-80 mesh, ≥99%
Sigma-Aldrich
Cadmium, granular, ≥99%, 5-20 mesh
Sigma-Aldrich
Cadmium, powder, −100 mesh, 99.5% trace metals basis
Sigma-Aldrich
Cadmium, shot, 3 mm, 99.999% trace metals basis
Cadmium, foil, not light tested, 50x50mm, thickness 0.005mm, 99.7+%
Cadmium, foil, not light tested, 25x25mm, thickness 0.01mm, 99.7+%
Cadmium, wire reel, 0.1m, diameter 1.0mm, hard, 99.999%
Cadmium, foil, 15mm disks, thickness 0.125mm, as rolled, 99.95%
Cadmium, wire reel, 0.5m, diameter 0.5mm, hard, 99.95+%
Cadmium, foil, 25mm disks, thickness 0.125mm, as rolled, 99.95%
Cadmium, foil, light tested, 25x25mm, thickness 0.05mm, as rolled, 99.99+%
Cadmium, foil, not light tested, 100x100mm, thickness 0.005mm, 99.7+%
Cadmium, foil, 8mm disks, thickness 0.125mm, as rolled, 99.99+%
Cadmium, foil, 6mm disks, thickness 0.005mm, 99.7+%
Cadmium, rod, 50mm, diameter 10.0mm, 99.999%
Cadmium, foil, not light tested, 25x25mm, thickness 0.002mm, 99.7+%
Cadmium, foil, 6mm disks, thickness 0.5mm, as rolled, 99.95%
Cadmium, foil, 6mm disks, thickness 0.25mm, as rolled, 99.95%
Cadmium, rod, 100mm, diameter 10.0mm, 99.999%
Cadmium, foil, light tested, 50x50mm, thickness 0.015mm, 99.7+%
Cadmium, foil, 8mm disks, thickness 0.01mm, 99.7+%
Cadmium, foil, not light tested, 100x100mm, thickness 0.015mm, 99.7+%
Cadmium, foil, 8mm disks, thickness 0.25mm, as rolled, 99.9%
Cadmium, foil, 6mm disks, thickness 0.015mm, 99.7+%
Cadmium, rod, 50mm, diameter 40mm, 99.9%
Cadmium, foil, 4mm disks, thickness 1.0mm, as rolled, 100%
Cadmium, foil, light tested, 25x25mm, thickness 0.015mm, 99.7+%
Cadmium, foil, 6mm disks, thickness 0.25mm, as rolled, 99.99+%
Cadmium, foil, 6mm disks, thickness 1.0mm, as rolled, 100%
Cadmium, rod, 1000mm, diameter 5.0mm, as drawn, 99.99+%