Merck
  • Phase-Transfer Catalysed Permanganate Oxidations Using Tris [2-(2-Methoxyethoxy) Ethyl] Amine (Tda-1).
Phase-Transfer Catalysed Permanganate Oxidations Using Tris [2-(2-Methoxyethoxy) Ethyl] Amine (Tda-1).
Synthetic Communications (1987)
Mckillop A and Mills LS.