Merck
  • PhLP3 modulates CCT-mediated actin and tubulin folding via ternary complexes with substrates.
PhLP3 modulates CCT-mediated actin and tubulin folding via ternary complexes with substrates.
The Journal of biological chemistry (2006-01-18)
Peter C Stirling, Jorge Cuéllar, Gabriel A Alfaro, Fatima El Khadali, Christopher T Beh, José M Valpuesta, Ronald Melki, Michel R Leroux
ABSTRACT
Many ATP-dependent molecular chaperones, including Hsp70, Hsp90, and the chaperonins GroEL/Hsp60, require cofactor proteins to regulate their ATPase activities and thus folding functions in vivo. One conspicuous exception has been the eukaryotic chaperonin CCT, for which no regulator of its ATPase activity, other than non-native substrate proteins, is known. We identify the evolutionarily conserved PhLP3 (phosducin-like protein 3) as a modulator of CCT function in vitro and in vivo. PhLP3 binds CCT, spanning the cylindrical chaperonin cavity and contacting at least two subunits. When present in a ternary complex with CCT and an actin or tubulin substrate, PhLP3 significantly diminishes the chaperonin ATPase activity, and accordingly, excess PhLP3 perturbs actin or tubulin folding in vitro. Most interestingly, however, the Saccharomyces cerevisiae PhLP3 homologue is required for proper actin and tubulin function. This cellular role of PhLP3 is most apparent in a strain that also lacks prefoldin, a chaperone that facilitates CCT-mediated actin and tubulin folding. We propose that the antagonistic actions of PhLP3 and prefoldin serve to modulate CCT activity and play a key role in establishing a functional cytoskeleton in vivo.
Sigma-Aldrich
Anti-CCT6B antibody produced in rabbit, affinity isolated antibody