Merck
  • Synthesis and evaluation of 2-(2-fluoro-4-hydroxymethyl-5-methoxy-phenoxy)acetic acid as a linker in solid-phase synthesis monitored by gel-phase (19)F NMR spectroscopy.
Synthesis and evaluation of 2-(2-fluoro-4-hydroxymethyl-5-methoxy-phenoxy)acetic acid as a linker in solid-phase synthesis monitored by gel-phase (19)F NMR spectroscopy.
Organic & biomolecular chemistry (2007-07-20)
Fredrik K Wallner, Sara Spjut, Dan Boström, Mikael Elofsson
ABSTRACT
Gel-phase (19)F NMR spectroscopy is a useful monitoring technique for solid-phase organic chemistry due to the high information content it delivers and swift acquisition times, using standard NMR spectrometers. This paper describes the synthesis of the novel linker 2-(2-fluoro-4-hydroxymethyl-5-methoxy-phenoxy)acetic acid in 29% yield over seven steps, using nucleophilic aromatic substitutions on 2,4,5-trifluorobenzonitrile as key steps. Following standard solid-phase synthesis a peptide could be cleaved from the linker using 20% TFA in CH(2)Cl(2) in 30 minutes, in contrast to a previously described monoalkoxy linker that requires 90% TFA in water at elevated temperature. A resin-bound peptide could be successfully glycosylated using only two equivalents of a thioglycoside donor, activated with N-iodosuccinimide and trifluoromethanesulfonic acid, and subsequent cleavage and deprotection gave the target glycopeptide. Direct glycosylation of the linker itself followed by mild acidic cleavage gave a fully protected hemiacetal for further chemical manipulation.