Merck
  • The protective effects of melittin on Propionibacterium acnes-induced inflammatory responses in vitro and in vivo.
The protective effects of melittin on Propionibacterium acnes-induced inflammatory responses in vitro and in vivo.
The Journal of investigative dermatology (2014-02-06)
Woo-Ram Lee, Kyung-Hyun Kim, Hyun-Jin An, Jung-Yeon Kim, Young-Chae Chang, Hyun Chung, Yoon-Yub Park, Myeong-Lyeol Lee, Kwan-Kyu Park
ABSTRACT
Melittin is the main component in the venom of the honey bee (Apis mellifera). It has multiple effects including antibacterial, antiviral, and anti-inflammatory activities in various cell types. However, the anti-inflammatory mechanisms of melittin have not been elucidated in Propionibactierium acnes (P. acnes)-induced keratinocyte or inflammatory skin disease animal models. In this study, we examined the effects of melittin on the production of inflammatory cytokines in heat-killed P. acnes-induced HaCaT cells. Heat-killed P. acnes-treated keratinocytes increased the expression of pro-inflammatory cytokines and Toll-like receptor 2. However, melittin treatment significantly suppressed the expression of these cytokines through regulation of the NF-κB and MAPK signaling pathways. Subsequently, the living P. acnes (1 × 10(7) CFU) were intradermally injected into the ear of mice. Living P. acnes-injected ears showed cutaneous erythema, swelling, and granulomatous response at 24 hours after injection. However, melittin-treated ears showed markedly reduced swelling and granulomatous responses compared with ears injected with only living P. acnes. These results demonstrate the feasibility of applying melittin for the prevention of inflammatory skin diseases induced by P. acnes.
Sigma-Aldrich
Melittin from honey bee venom, ≥65% (HPLC)
Sigma-Aldrich
Melittin, ≥97% (HPLC), synthetic
Sigma-Aldrich
Melittin from honey bee venom, ≥85% (HPLC)