Merck
  • Spontaneous reductions in smoking during double-blind buprenorphine detoxification.
Spontaneous reductions in smoking during double-blind buprenorphine detoxification.
Addictive behaviors (2014-05-23)
Mollie E Patrick, Kelly E Dunn, Gary J Badger, Sarah H Heil, Stephen T Higgins, Stacey C Sigmon
ABSTRACT
Evidence suggests a positive association between administration of psychoactive drugs and rates of cigarette smoking. Prevalence of smoking among opioid-dependent individuals, for example, is four times greater than the general population. We recently completed a randomized double-blind trial evaluating outpatient buprenorphine taper for prescription opioid (PO) abusers, which provided a unique opportunity to examine naturalistic changes in smoking among participants who detoxified without resumption of illicit opioid use. Participants received no smoking-cessation services and were not encouraged to alter their smoking in any way. A subset of 10 opioid-dependent smokers, who were randomized to receive the same 4-week buprenorphine taper and successfully completed detoxification, were included in the present study. They provided staff-observed urine specimens thrice-weekly throughout the 12-week trial. Specimens were analyzed on-site via enzyme-multiplied immunoassay for urinary cotinine, a metabolite of nicotine that provides a sensitive biochemical measure of smoking status. Mean cotinine levels were significantly different across study phases, with significantly lower cotinine levels during taper (1317.5 ng/ml) and post-taper (1015.8 ng/ml) vs. intake (1648.5 ng/ml) phases (p''s<.05). Overall, mean cotinine levels decreased by 38% between intake and end-of-study, reflecting a reduction of approximately eight cigarettes per day. These data provide additional evidence that opioids influence smoking and extend prior findings to include primary PO abusers, rigorous double-blind opioid dosing conditions and urinary cotinine. These results also suggest that, while likely insufficient for complete cessation, patients who successfully taper from opioids may also experience concurrent reductions in smoking and thus may be ideal candidates for smoking cessation services.
Product Description
Supelco
Buprenorphine solution, 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
(−)-Cotinine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Buprenorphine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Buprenorphine hydrochloride solution, analytical standard, for drug analysis
Sigma-Aldrich
Buprenorphine hydrochloride
Sigma-Aldrich
(−)-Cotinine, ≥98%