Merck
  • Whole-body phenylalanine kinetics and skeletal muscle protein signaling in horses with pituitary pars intermedia dysfunction.
Whole-body phenylalanine kinetics and skeletal muscle protein signaling in horses with pituitary pars intermedia dysfunction.
American journal of veterinary research (2014-06-25)
Laurel M Mastro, Amanda A Adams, Kristine L Urschel
ABSTRACT
To compare whole-body phenylalanine kinetics and the abundance of factors in signaling pathways associated with skeletal muscle protein synthesis and protein breakdown between horses with pituitary pars intermedia dysfunction (PPID) and age-matched control horses without PPID. 12 aged horses (6 horses with PPID and 6 control horses; mean age, 25.0 and 25.7 years, respectively). Plasma glucose, insulin, and amino acids concentrations were determined before and 90 minutes after feeding. Gluteal muscle biopsy samples were obtained from horses 90 minutes after feeding, and the abundance and activation of factors involved in signaling pathways of muscle protein synthesis and breakdown were determined. The next day, horses received a priming dose and 2 hours of a constant rate infusion of (13)C sodium bicarbonate followed by a priming dose and 4 hours of a constant rate infusion of 1-(13)C phenylalanine IV; whole-body protein synthesis was determined. Plasma glucose and insulin concentrations were higher after feeding than they were before feeding for both groups of horses; however, no significant postprandial increase in plasma amino acids concentrations was detected for either group. Phenylalanine flux, oxidation, release from protein breakdown, and nonoxidative disposal were not significantly different between groups. No significant effect of PPID status was detected on the abundance or activation of positive or negative regulators of protein synthesis or positive regulators of protein breakdown. Results of this study suggested that whole-body phenylalanine kinetics and the postprandial activation of signaling pathways that regulate protein synthesis and breakdown in muscles were not affected by PPID status alone in aged horses.
Product Description
Supelco
L-Threonine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Sodium bicarbonate, tested according to Ph. Eur.
Sigma-Aldrich
L-Threonine, BioXtra, ≥99.5% (NT)
SAFC
L-Threonine
Sigma-Aldrich
DL-Tryptophan, ≥99% (HPLC)
Sigma-Aldrich
DL-Histidine, ≥99% (TLC)
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
L-Threonine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
L-Threonine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
DL-Tryptophan, ≥99% (HPLC)
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, Molecular Biology, suitable for cell culture, suitable for insect cell culture
Leucine, European Pharmacopoeia (EP) Reference Standard
Isoleucine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Supelco
L-Threonine, Pharmaceutical Secondary Standard; Certified Reference Material
Tryptophan, European Pharmacopoeia (EP) Reference Standard
Valine, European Pharmacopoeia (EP) Reference Standard
L-Threonine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium bicarbonate, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium bicarbonate, ReagentPlus®, ≥99.5%, powder
Sigma-Aldrich
Sodium bicarbonate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E500, 99.0-100.5%, powder
USP
Sodium bicarbonate, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium hydrogencarbonate, −40-+140 mesh, ≥95%
Sigma-Aldrich
DL-Phenylalanine, ReagentPlus®, 99%
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C
Sigma-Aldrich
DL-Valine, ReagentPlus®, ≥99.0% (NT)
Sigma-Aldrich
L-Isoleucine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
DL-Valine, ≥97%
Sigma-Aldrich
L-Phenylalanine, 99%, FCC