Merck
  • High-dose chemotherapy plus autologous stem-cell transplantation as consolidation therapy in patients with relapsed multiple myeloma after previous autologous stem-cell transplantation (NCRI Myeloma X Relapse [Intensive trial]): a randomised, open-label, phase 3 trial.
High-dose chemotherapy plus autologous stem-cell transplantation as consolidation therapy in patients with relapsed multiple myeloma after previous autologous stem-cell transplantation (NCRI Myeloma X Relapse [Intensive trial]): a randomised, open-label, phase 3 trial.
The Lancet. Oncology (2014-06-21)
Gordon Cook, Cathy Williams, Julia M Brown, David A Cairns, Jamie Cavenagh, John A Snowden, A John Ashcroft, Marie Fletcher, Chris Parrish, Kwee Yong, Jim Cavet, Hanna Hunter, Jenny M Bird, Anna Chalmers, Sheila O'Connor, Mark T Drayson, Treen C M Morris
ABSTRACT
Relapsed multiple myeloma has no standard treatment, and the role of autologous stem-cell transplantation (ASCT) has not been fully defined. We aimed to compare high-dose melphalan plus salvage ASCT with cyclophosphamide in patients with relapsed multiple myeloma who had previously undergone ASCT. This multicentre, randomised, open-label, phase 3 study recruited patients aged at least 18 years with multiple myeloma who needed treatment for first progressive or relapsed disease at least 18 months after a previous ASCT from 51 centres across the UK. Before randomisation, eligible patients received bortezomib, doxorubicin, and dexamethasone (PAD) induction therapy and then underwent peripheral blood stem-cell mobilisation and harvesting if applicable. Eligible patients (with adequate stem-cell harvest) were randomly assigned (1:1), using an automated telephone randomisation line, to either high-dose melphalan 200 mg/m(2) plus salvage ASCT or oral cyclophosphamide (400mg/m(2) per week for 12 weeks). Randomisation was stratified by length of first remission or plateau and response to PAD re-induction therapy. The primary endpoint was time to disease progression, analysed by intention to treat. This trial is registered with ClinicalTrials.gov, number NCT00747877, and EudraCT, number 2006-005890-24. Between April 16, 2008, and Nov 19, 2012, 297 patients were registered, of whom 293 received PAD re-induction therapy. Between Aug 26, 2008, and Nov 16, 2012, 174 patients with sufficient PBSCs were randomised to salvage ASCT (n=89) or cyclophosphamide (n=85). After a median follow-up of 31 months (IQR 19-42), median time to progression was significantly longer in the salvage ASCT than in the cyclophosphamide group (19 months [95% CI 16-25] vs 11 months [9-12]; hazard ratio 0·36 [95% CI 0·25-0·53]; p<0·0001). Frequently reported (in >10% of patients) grade 3-4 adverse events with PAD induction, salvage ASCT, and cyclophosphamide were: neutropenia (125 [43%] of 293 patients after PAD, and 63 [76%] of 83 patients in the salvage ASCT group vs 11 [13%] of 84 patients in the cyclophosphamide group), thrombocytopenia (150 [51%] after PAD, and 60 [72%] vs four [5%], respectively), and peripheral neuropathy (35 [12%] after PAD, and none vs none, respectively). This study provides evidence for the improved efficacy of high-dose melphalan plus salvage ASCT when compared with cyclophosphamide in patients with relapsed multiple myeloma eligible for intensive therapy, which might help to guide clinical decisions regarding the management of such patients. Cancer Research UK.
Sigma-Aldrich
Doxorubicin hydrochloride, suitable for fluorescence, 98.0-102.0% (HPLC)
Sigma-Aldrich
Doxorubicin hydrochloride, 98.0-102.0% (HPLC)
Doxorubicin hydrochloride, European Pharmacopoeia (EP) Reference Standard
Supelco
Dexamethasone solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Dexamethasone for peak identification, European Pharmacopoeia (EP) Reference Standard
USP
Doxorubicin hydrochloride, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dexamethasone-Water Soluble, BioReagent, suitable for cell culture
Sigma-Aldrich
Dexamethasone, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Dexamethasone, meets USP testing specifications
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder
Sigma-Aldrich
Melphalan, powder
Supelco
Dexamethasone, VETRANAL®, analytical standard
Sigma-Aldrich
Dexamethasone, tested according to Ph. Eur.
Dexamethasone, European Pharmacopoeia (EP) Reference Standard
Supelco
Dexamethasone, Pharmaceutical Secondary Standard; Certified Reference Material
Dexamethasone for system suitability, European Pharmacopoeia (EP) Reference Standard