Merck
  • Effect of atorvastatin on low-density lipoprotein subpopulations and comparison between indicators of plasma atherogenicity: a pilot study.
Effect of atorvastatin on low-density lipoprotein subpopulations and comparison between indicators of plasma atherogenicity: a pilot study.
Angiology (2013-10-29)
Marek Kucera, Stanislav Oravec, Eva Hirnerova, Nada Huckova, Zuzana Celecova, Ludovit Gaspar, Maciej Banach
ABSTRACT
Treatment with statins to achieve target low-density lipoprotein cholesterol (LDL-C) levels is still associated with residual risk. Lipoprotein subfraction evaluation can provide additional information regarding atherogenicity in these individuals. Patients (n = 40) with hypercholesterolemia (29 females, mean age 63 years), without previous hypolipemic treatment, were treated with atorvastatin 40 mg/d for 3 months. Atorvastatin significantly reduced total cholesterol (6.7 ± 1.0 vs 4.6 ± 1.3 mmol/L, P < .001), LDL-C (4.3 ± 1.0 vs 2.6 ± 0.9 mmol/L, P < .001), triglycerides (1.8 ± 0.9 vs 1.5 ± 1.00 mmol/L, P < .05), small-dense LDL (sdLDL) fraction 3 to 7 (0.22 ± 0.37 vs 0.09 ± 0.16 mmol/L, P < .001), and apolipoprotein B (apoB; 1.0 ± 0.2 vs 0.74 ± 0.2 g/L, P < .001). There was a negative correlation of atherogenic index of plasma (AIP) with buoyant LDL-1 and LDL-2 (r = -.35; P < .05) and positive with sdLDL-3 to sdLDL-7 (r = .52, P < .001). Administration of atorvastatin 40 mg/d in patients with hypercholesterolemia caused a shift in sdLDL subfractions to large, buoyant subfractions. The AIP better correlated with sdLDL than apoB levels.
Product Description
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Cholesterol, tested according to Ph. Eur.
Sigma-Aldrich
Cholesterol, from lanolin, ≥99.0% (GC)
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder