Merck
  • Cell cycle arrest and apoptosis induced by 1α,25(OH)2D3 and TX 527 in Kaposi sarcoma is VDR dependent.
Cell cycle arrest and apoptosis induced by 1α,25(OH)2D3 and TX 527 in Kaposi sarcoma is VDR dependent.
The Journal of steroid biochemistry and molecular biology (2013-12-10)
Verónica González-Pardo, Alejandra Suares, Annemieke Verstuyf, Pierre De Clercq, Ricardo Boland, Ana Russo de Boland
ABSTRACT
We have previously shown that 1α,25(OH)2-Vitamin D3 [1α,25(OH)2D3] and its less calcemic analog TX 527 inhibit the proliferation of endothelial cells transformed by the viral G protein-coupled receptor associated to Kaposi sarcoma (vGPCR) and this could be partially explained by the inhibition of the NF-κB pathway. In this work, we further explored the mechanism of action of both vitamin D compounds in Kaposi sarcoma. We investigated whether the cell cycle arrest and subsequent apoptosis of endothelial cells (SVEC) and SVEC transformed by vGPCR (SVEC-vGPCR) elicited by 1α,25(OH)2D3 and TX 527 were mediated by the vitamin D receptor (VDR). Cell cycle analysis of SVEC and SVEC-vGPCR treated with 1α,25(OH)2D3 (10nM, 48h) revealed that 1α,25(OH)2D3 increased the percentage of cells in the G0/G1 phase and diminished the percentage of cells in the S phase of the cell cycle. Moreover, the number of cells in the S phase was higher in SVEC-vGPCR than in SVEC due to vGPCR expression. TX 527 exerted similar effects on growth arrest in SVEC-vGPCR cells. The cell cycle changes were suppressed when the expression of the VDR was blocked by a stable transfection of shRNA against VDR. Annexin V-PI staining demonstrated apoptosis in both SVEC and SVEC-vGPCR after 1α,25(OH)2D3 and TX 527 treatment (10nM, 24h). Cleavage of caspase-3 detected by Western blot analysis was increased to a greater extent in SVEC than in SVEC-vGPCR cells, and this effect was also blocked in VDR knockdown cells. Altogether, these results suggest that 1α,25(OH)2D3 and TX 527 inhibit the proliferation of SVEC and SVEC-vGPCR and induce apoptosis by a mechanism that involves the VDR.
Product Description
USP
Cholecalciferol, United States Pharmacopeia (USP) Reference Standard
Cholecalciferol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Cholecalciferol, analytical standard
Supelco
Cholecalciferol (D3), analytical standard
Sigma-Aldrich
Cholecalciferol, meets USP testing specifications
Sigma-Aldrich
Cholecalciferol, ≥98% (HPLC)
Calcitriol, European Pharmacopoeia (EP) Reference Standard
Supelco
1α,25-Dihydroxyvitamin D3 solution, 5 μg/mL in ethanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
1α,25-Dihydroxyvitamin D3, ≥97.0% (HPLC)
Sigma-Aldrich
1α,25-Dihydroxyvitamin D3, ≥99% (HPLC)
Supelco
Cholecalciferol (Vitamin D3), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Vitamin D3 solution, 1 mg/mL in ethanol, ampule of 1 mL, certified reference material, Cerilliant®