Merck
  • Evaluation of corneal stromal demarcation line depth following standard and a modified-accelerated collagen cross-linking protocol.
Evaluation of corneal stromal demarcation line depth following standard and a modified-accelerated collagen cross-linking protocol.
American journal of ophthalmology (2014-07-19)
George D Kymionis, Konstantinos I Tsoulnaras, Michael A Grentzelos, Dimitrios A Liakopoulos, Nikolaos G Tsakalis, Styliani V Blazaki, Theodoros A Paraskevopoulos, Miltiadis K Tsilimbaris
ABSTRACT
To compare the corneal stromal demarcation line depth using anterior segment optical coherence tomography (AS-OCT) after corneal collagen cross-linking (CXL) using 2 different treatment protocols: the standard Dresden protocol (30 minutes with 3 mW/cm(2)) and a modified-accelerated protocol (14 minutes with 9 mW/cm(2)). Prospective, comparative study. Forty-three keratoconic patients (52 eyes) were enrolled. All patients underwent CXL using the same high-intensity ultraviolet-A (UV-A) irradiation device. Twenty-six eyes were treated for 30 minutes with 3 mW/cm(2) according to the standard Dresden protocol (Group 1), while 26 eyes were treated with a novel modified-accelerated CXL protocol for 14 minutes with 9 mW/cm(2) of UV-A irradiation intensity (Group 2). One month postoperatively, corneal stromal demarcation line depth was measured by 2 independent observers using AS-OCT. Corneal stromal demarcation line depth was assessed with no significant difference between observer measurements for both groups (P = .676 for Group 1 and P = .566 for Group 2). Mean corneal stromal demarcation line depth was 337.00 ± 46.46 μm for Group 1 and 322.91 ± 48.28 μm for Group 2. There was no statistically significant difference (P = .243) in the corneal stromal demarcation line depth between the 2 groups. Corneal stromal demarcation line depth using UV-A with 3 mW/cm(2) for 30 minutes and 9 mW/cm(2) for 14 minutes was similar. A modified-accelerated protocol of 14 minutes of CXL provided the same treatment depth as the classic Dresden protocol.
Product Description
Riboflavin, European Pharmacopoeia (EP) Reference Standard
USP
Riboflavin, United States Pharmacopeia (USP) Reference Standard
Supelco
Riboflavin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Collagen, Type I solution from rat tail, BioReagent, suitable for cell culture, sterile-filtered
Sigma-Aldrich
Collagen from rat tail, Bornstein and Traub Type I, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type V (Sigma Type IX), powder
Sigma-Aldrich
Collagen from bovine tracheal cartilage, Bornstein and Traub Type II, powder
Sigma-Aldrich
Collagen from bovine nasal septum, Bornstein and Traub Type II, powder
Sigma-Aldrich
Collagen from Engelbreth-Holm-Swarm murine sarcoma basement membrane, Type IV (Miller), lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type III (Sigma Type X), powder
Sigma-Aldrich
Collagen from rabbit skin, Bornstein and Traub Type I, powder
Sigma-Aldrich
(−)-Riboflavin, meets USP testing specifications
Sigma-Aldrich
(−)-Riboflavin, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98%
Sigma-Aldrich
(−)-Riboflavin, from Eremothecium ashbyii, ≥98%
Sigma-Aldrich
Collagen Type IV from human cell culture, Bornstein and Traub Type IV, 0.3 mg/mL, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from chicken sternal cartilage, Type II (Miller), powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen human, Bornstein and Traub Type I, acid soluble, powder, ~95% (SDS-PAGE)
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type I (Sigma Type VIII), powder
Sigma-Aldrich
Collagen from bovine achilles tendon, powder, suitable for substrate for collagenase
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I, (0.1% solution in 0.1 M acetic acid), aseptically processed, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I, solid, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder
Supelco
Riboflavin (B2), analytical standard
Riboflavin for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, solution, suitable for cell culture, High Performance