Merck
  • Hajek revisited: a histological examination of the quadrangular membrane.
Hajek revisited: a histological examination of the quadrangular membrane.
The Annals of otology, rhinology, and laryngology (2014-06-21)
Nwanmegha Young, Mikhail Wadie Abdelmessih, Clarence Sasaki
ABSTRACT
The predictability of laryngeal cancer spread is due in part to connective tissue membranes. These membranes function as barriers to cancer and divide the larynx into subunits. The field of laryngeal conservation surgery is based on these concepts. The quadrangular membrane plays an important role, hindering the lateral spread of cancer in the larynx. The composition of this membrane has not been well described in the literature. In this study, we examine basic characteristics of the quadrangular membrane using histological techniques. Whole organ sections of the larynx were used. These sections were examined under a microscope with stains specific for collagen and elastin. Examination of the sections revealed that the quadrangular membrane is made up of closely woven undulating collagen and elastic fibers. The quadrangular membrane is a fibroelastic structure providing a barrier to cancer spread.
Sigma-Aldrich
Collagen Type IV from human cell culture, Bornstein and Traub Type IV, 0.3 mg/mL, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type I (Sigma Type VIII), powder
Sigma-Aldrich
Collagen from bovine achilles tendon, powder, suitable for substrate for collagenase
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I, solid, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I, (0.1% solution in 0.1 M acetic acid), aseptically processed, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from chicken sternal cartilage, Type II (Miller), powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen human, Bornstein and Traub Type I, acid soluble, powder, ~95% (SDS-PAGE)
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, solution, suitable for cell culture, High Performance
Sigma-Aldrich
Collagen, Type I solution from rat tail, BioReagent, suitable for cell culture, sterile-filtered
Sigma-Aldrich
Collagen from rabbit skin, Bornstein and Traub Type I, powder
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type III (Sigma Type X), powder
Sigma-Aldrich
Collagen from Engelbreth-Holm-Swarm murine sarcoma basement membrane, Type IV (Miller), lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from bovine tracheal cartilage, Bornstein and Traub Type II, powder
Sigma-Aldrich
Collagen from bovine nasal septum, Bornstein and Traub Type II, powder
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type V (Sigma Type IX), powder
Sigma-Aldrich
Collagen from rat tail, Bornstein and Traub Type I, powder, BioReagent, suitable for cell culture