Merck
  • Concise Synthesis of Sibrafiban and Lamifiban, Two Non-Peptide Fibrinogen Receptor (GPIIb/IIIa) Antagonists.
Concise Synthesis of Sibrafiban and Lamifiban, Two Non-Peptide Fibrinogen Receptor (GPIIb/IIIa) Antagonists.
J. Chin. Chem. Soc. (2001)
Chang MY and Chen ST.