Merck
  • A simple one-pot preparation of diazoacetoacetate derivatives from aldehydes.
A simple one-pot preparation of diazoacetoacetate derivatives from aldehydes.
The Journal of organic chemistry (2008-10-09)
Marvis O Erhunmwunse, Patrick G Steel
ABSTRACT
Diazoacetoacetate derivatives can be simply and efficiently prepared from aldehydes in a one-pot process involving initial DBU-promoted "aldol" condensation with ethyl diazoacetate followed by in situ oxidation with IBX. Aryl, alkyl, and unsaturated aldehydes are all viable substrates.