Merck
  • Assessment of cytochrome C oxidase dysfunction in the substantia nigra/ventral tegmental area in schizophrenia.
Assessment of cytochrome C oxidase dysfunction in the substantia nigra/ventral tegmental area in schizophrenia.
PloS one (2014-06-19)
Matthew W Rice, Kristen L Smith, Rosalinda C Roberts, Emma Perez-Costas, Miguel Melendez-Ferro
ABSTRACT
Perturbations in metabolism are a well-documented but complex facet of schizophrenia pathology. Optimal cellular performance requires the proper functioning of the electron transport chain, which is constituted by four enzymes located within the inner membrane of mitochondria. These enzymes create a proton gradient that is used to power the enzyme ATP synthase, producing ATP, which is crucial for the maintenance of cellular functioning. Anomalies in a single enzyme of the electron transport chain are sufficient to cause disruption of cellular metabolism. The last of these complexes is the cytochrome c oxidase (COX) enzyme, which is composed of thirteen different subunits. COX is a major site for oxidative phosphorylation, and anomalies in this enzyme are one of the most frequent causes of mitochondrial pathology. The objective of the present report was to assess if metabolic anomalies linked to COX dysfunction may contribute to substantia nigra/ventral tegmental area (SN/VTA) pathology in schizophrenia. We tested COX activity in postmortem SN/VTA from schizophrenia and non-psychiatric controls. We also tested the protein expression of key subunits for the assembly and activity of the enzyme, and the effect of antipsychotic medication on subunit expression. COX activity was not significantly different between schizophrenia and non-psychiatric controls. However, we found significant decreases in the expression of subunits II and IV-I of COX in schizophrenia. Interestingly, these decreases were observed in samples containing the entire rostro-caudal extent of the SN/VTA, while no significant differences were observed for samples containing only mid-caudal regions of the SN/VTA. Finally, rats chronically treated with antipsychotic drugs did not show significant changes in COX subunit expression. These findings suggest that COX subunit expression may be compromised in specific sub-regions of the SN/VTA (i.e. rostral regions), which may lead to a faulty assembly of the enzyme and a greater vulnerability to metabolic insult.
Sigma-Aldrich
Sucrose, meets USP testing specifications
USP
Sucrose, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sucrose, European Pharmacopoeia (EP) Reference Standard
Supelco
Sucrose, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Cytochrome c from bovine heart, ≥95% based on Mol. Wt. 12,327 basis, powder, suitable for mammalian cell culture
Sigma-Aldrich
Cytochrome c Oxidase from bovine heart, 5 mg protein/mL
Sigma-Aldrich
3,3′-Diaminobenzidine tetrahydrochloride hydrate, ≥96%
Sigma-Aldrich
Sucrose, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioXtra
Supelco
Sucrose, analytical standard, for enzymatic assay kit SCA20
Sigma-Aldrich
Sucrose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Millipore
Sucrose, suitable for microbiology, ACS reagent, ≥99.0%
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Sucrose, ≥99% (GC), Grade I, suitable for plant cell culture
Sigma-Aldrich
Sucrose, BioUltra, Molecular Biology, ≥99.5% (HPLC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Anti-Actin Antibody, clone C4, ascites fluid, clone C4, Chemicon®
Sigma-Aldrich
4-Nitrophenyl phosphate disodium salt hexahydrate, suitable for enzyme immunoassay, ≥99.0% (enzymatic)
Sigma-Aldrich
Phosphatase substrate, 40 mg capsules
Sigma-Aldrich
Phosphatase substrate, 100 mg capsules
Sigma-Aldrich
Phosphatase substrate, powder
Sigma-Aldrich
Phosphatase substrate, 40 mg tablets
Sigma-Aldrich
Phosphatase substrate, 5 mg tablets
Sigma-Aldrich
4-Nitrophenyl phosphate disodium salt hexahydrate, tablet
Sigma-Aldrich
4-Nitrophenyl phosphate disodium salt hexahydrate, tablet
Sigma-Aldrich
4-Nitrophenyl phosphate disodium salt hexahydrate, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
4-Nitrophenyl phosphate disodium salt hexahydrate, tablet