Merck
  • The impact of prior authorization on buprenorphine dose, relapse rates, and cost for Massachusetts Medicaid beneficiaries with opioid dependence.
The impact of prior authorization on buprenorphine dose, relapse rates, and cost for Massachusetts Medicaid beneficiaries with opioid dependence.
Health services research (2014-07-22)
Robin E Clark, Jeffrey D Baxter, Bruce A Barton, Gideon Aweh, Elizabeth O'Connell, William H Fisher
ABSTRACT
To assess the impact of a 2008 dose-based prior authorization policy for Massachusetts Medicaid beneficiaries using buprenorphine + naloxone for opioid addiction treatment. Doses higher than 16 mg required progressively more frequent authorizations. Mediciaid claims for 2007 and 2008 linked with Department of Public Health (DPH) service records. We conducted time series for all buprenorphine users and a longitudinal cohort analysis of 2,049 individuals who began buprenorphine treatment in 2007. Outcome measures included use of relapse-related services, health care expenditures per person, and buprenorphine expenditures. We used ICD-9 codes and National Drug Codes to identify individuals with opioid dependence who filled prescriptions for buprenorphine. Medicaid and DPH data were linked with individual identifiers. Individuals using doses >24 mg decreased from 16.5 to 4.1 percent. Relapses increased temporarily for some users but returned to previous levels within 3 months. Buprenorphine expenditures decreased but total expenditures did not change significantly. Prior authorization policies strategically targeted by dose level appear to successfully reduce use of higher than recommended buprenorphine doses. Savings from these policies are modest and may be accompanied by brief increases in relapse rates. Lower doses may decrease diversion of buprenorphine.
Sigma-Aldrich
Buprenorphine hydrochloride
Supelco
Buprenorphine hydrochloride solution, analytical standard, for drug analysis
Supelco
Buprenorphine solution, 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Buprenorphine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®