Merck
  • Effect of low-concentration povidone iodine on postoperative complications after third molar surgery: a pilot split-mouth study.
Effect of low-concentration povidone iodine on postoperative complications after third molar surgery: a pilot split-mouth study.
Journal of oral and maxillofacial surgery : official journal of the American Association of Oral and Maxillofacial Surgeons (2014-09-25)
Hamid Mahmoud Hashemi, Farnoosh Mohammadi, Mahboube Hasheminasab, Ali Mahmoud Hashemi, Shohreh Zahraei, Talieh Mahmoud Hashemi
ABSTRACT
Povidone iodine is used primarily as an antiseptic to decrease surgical site infection. Its hemostatic and antiedematous properties in oral surgery also have been investigated recently. A randomized controlled clinical trial was performed in 30 patients undergoing mandibular third molar removal in a split-mouth design. In the study group, a povidone iodine solution with a concentration of 0.5 mg/mL was used as the coolant and irrigant solution, whereas normal saline was used in the control group. Swelling (orotragus and mentotragus distances), trismus (maximum interincisal opening), and pain (visual analog scale score) were evaluated on postoperative days 2 and 7. In the study group, a significant decrease in swelling and trismus was observed at the 2 postoperative visits (P = .00) compared with the control group. The decrease of pain in the study group was not statistically significant at either postoperative visit (P > .05). More patients (63%) were subjectively satisfied with the side treated with povidone iodine. Povidone iodine irrigation is an inexpensive and safe method to lessen the postoperative sequelae of third molar surgery.
Sigma-Aldrich
Poly(vinylpyrrolidone)–Iodine complex
Povidone (iodinated), European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)