Merck
  • Primary care-based buprenorphine taper vs maintenance therapy for prescription opioid dependence: a randomized clinical trial.
Primary care-based buprenorphine taper vs maintenance therapy for prescription opioid dependence: a randomized clinical trial.
JAMA internal medicine (2014-10-21)
David A Fiellin, Richard S Schottenfeld, Christopher J Cutter, Brent A Moore, Declan T Barry, Patrick G O'Connor
ABSTRACT
Prescription opioid dependence is increasing and creates a significant public health burden, but primary care physicians lack evidence-based guidelines to decide between tapering doses followed by discontinuation of buprenorphine hydrochloride and naloxone hydrochloride therapy (hereinafter referred to as buprenorphine therapy) or ongoing maintenance therapy. To determine the efficacy of buprenorphine taper vs ongoing maintenance therapy in primary care-based treatment for prescription opioid dependence. We conducted a 14-week randomized clinical trial that enrolled 113 patients with prescription opioid dependence from February 17, 2009, through February 1, 2013, in a single primary care site. Patients were randomized to buprenorphine taper (taper condition) or ongoing buprenorphine maintenance therapy (maintenance condition). The buprenorphine taper was initiated after 6 weeks of stabilization, lasted for 3 weeks, and included medications for opioid withdrawal, after which patients were offered naltrexone treatment. The maintenance group received ongoing buprenorphine therapy. All patients received physician and nurse support and drug counseling. Illicit opioid use via results of urinanalysis and patient report, treatment retention, and reinitiation of buprenorphine therapy (taper group only). During the trial, the mean percentage of urine samples negative for opioids was lower for patients in the taper group (35.2% [95% CI, 26.2%-44.2%]) compared with those in the maintenance group (53.2% [95% CI, 44.3%-62.0%]). Patients in the taper group reported more days per week of illicit opioid use than those in the maintenance group once they were no longer receiving buprenorphine (mean use, 1.27 [95% CI, 0.60-1.94] vs 0.47 [95% CI, 0.19-0.74] days). Patients in the taper group had fewer maximum consecutive weeks of opioid abstinence compared with those in the maintenance group (mean abstinence, 2.70 [95% CI, 1.72-3.75] vs 5.20 [95% CI, 4.16-6.20] weeks). Patients in the taper group were less likely to complete the trial (6 of 57 [11%] vs 37 of 56 [66%]; P < .001). Sixteen patients in the taper group reinitiated buprenorphine treatment after the taper owing to relapse. Tapering is less efficacious than ongoing maintenance treatment in patients with prescription opioid dependence who receive buprenorphine therapy in primary care. clinicaltrials.gov Identifier: NCT00555425.
Product Description
Supelco
Buprenorphine solution, 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Buprenorphine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Buprenorphine hydrochloride
Supelco
Buprenorphine hydrochloride solution, analytical standard, for drug analysis
USP
Naloxone, United States Pharmacopeia (USP) Reference Standard