Merck
  • Identification and initial characterisation of a Plasmodium falciparum Cox17 copper metallochaperone.
Identification and initial characterisation of a Plasmodium falciparum Cox17 copper metallochaperone.
Experimental parasitology (2014-12-03)
David L Choveaux, Robert G E Krause, Jude M Przyborski, J P Dean Goldring
ABSTRACT
Copper is an essential micronutrient for all living organisms as an important catalytic co-factor for key enzymes. In higher eukaryotes intracellular copper is distributed by copper metallochaperones. Copper chelators such as neocuproine and tetrathiomolybdate inhibit Plasmodium falciparum erythrocytic development, indicating a requirement for copper by the parasite. A screen of the P. falciparum genome database identified eight potential copper-requiring protein orthologs, including four candidate copper metallochaperones implicated in the delivery of copper to cytochrome-c oxidase. A P. falciparum Cox17 ortholog (PfCox17) was recombinantly expressed and the purified protein bound reduced copper in vitro. PfCox17 was localised to the parasite cytoplasm. Characterisation of plasmodial proteins involved in copper metabolism will help us understand the role of this essential microelement in plasmodial homeostasis.
Product Description
Supelco
Ascorbic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
Ascorbic acid, European Pharmacopoeia (EP) Reference Standard
USP
Ascorbic acid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%
Copper (S, P), BCR®, certified reference material, chips
Copper, IRMM®, certified reference material, 0.1 mm foil
Copper, IRMM®, certified reference material, 0.5 mm wire
Sigma-Aldrich
Copper, ACS reagent, granular, 10-40 mesh, ≥99.90%
Copper, wire reel, 50m, diameter 0.5mm, as drawn, 99.98+%
Copper, wire reel, 20m, diameter 0.25mm, annealed, 99.9%
Copper, tube, 200mm, outside diameter 1.75mm, inside diameter 0.33mm, wall thickness 0.71mm, as drawn, 99.9%
Sigma-Aldrich
Copper, shot, −3-+14 mesh, 99%
Sigma-Aldrich
Copper, foil, thickness 0.25 mm, 99.98% trace metals basis
Sigma-Aldrich
Copper, powder (spheroidal), 10-25 μm, 98%
Sigma-Aldrich
Copper, powder, 99.99% trace metals basis
Sigma-Aldrich
Copper, nanopowder, 40-60 nm particle size (SAXS), ≥99.5% trace metals basis
Sigma-Aldrich
Copper, nanopowder, 60-80 nm particle size (APS), ≥99.5% trace metals basis
Sigma-Aldrich
Copper, powder (dendritic), <45 μm, 99.7% trace metals basis
Copper - O.F.H.C., foil, 300x300mm, thickness 0.25mm, annealed, 99.95+%
Copper - O.F.H.C., foil, 100x100mm, thickness 0.1mm, hard, 99.95+%
Copper, mesh, 100x100mm, nominal aperture 0.14mm, thickness 0.25mm, wire diameter 0.115mm, 100x100 wires/inch, open area 30.3%, plain weave mesh
Copper, wire reel, 1000m, diameter 0.25mm, annealed, 99.9%
Copper - O.F.H.C., rod, 1000mm, diameter 12.7mm, as drawn, 99.95+%
Sigma-Aldrich
Copper, powder, <425 μm, 99.5% trace metals basis
Copper, rod, 100mm, diameter 2.0mm, as drawn, 99.99+%
Copper, wire reel, 1000m, diameter 0.04mm, annealed, 99.9%
Copper, rod, 50mm, diameter 5.0mm, as drawn, 99.999%
Copper - O.F.H.C., wire reel, 2m, diameter 2.0mm, hard, 99.95+%