Merck
  • Evaluation of corneal changes after conventional versus accelerated corneal cross-linking: a randomized controlled trial.
Evaluation of corneal changes after conventional versus accelerated corneal cross-linking: a randomized controlled trial.
Journal of refractive surgery (Thorofare, N.J. : 1995) (2014-12-02)
Hesam Hashemian, Mahmoud Jabbarvand, Mehdi Khodaparast, Kambiz Ameli
ABSTRACT
To evaluate the outcome of accelerated and conventional corneal cross-linking (CXL) procedures regarding their effect on morphological and optical properties of the cornea. One hundred fifty-three eyes of 153 patients were evaluated before and during a 15-month follow-up period after CXL (76 eyes in the conventional group and 77 eyes in the accelerated group). Measured variables include corrected distance visual acuity (CDVA), uncorrected distance visual acuity (UDVA), refraction, maximum keratometry, endothelial cell density, anterior and posterior stromal keratocyte density, and subbasal nerve density. All variables were compared between the two study groups. Cylindrical and spherical components of refraction improved significantly during 15 months of follow-up. No difference was observed between the two study groups. UDVA and CDVA improved in the same manner, with no intergroup differences. Endothelial cell density did not change significantly during the follow-up period in either group. K-max increased slightly in the first month of the follow-up, but started to decrease at postoperative visits without any significant difference in the two groups. Anterior stromal keratocyte density and subbasal nerve density decreased significantly in both groups 1 month postoperatively. Both variables had a more significant decrease in the conventional group at all visits before the 1-year visit. At the final 15-month visit, there were no significant differences in any value between the two groups. Accelerated and conventional CXL seem to have a comparable and acceptable effect on keratoconus in the short-term follow-up period.
Sigma-Aldrich
Collagen Type IV from human cell culture, Bornstein and Traub Type IV, 0.3 mg/mL, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type I (Sigma Type VIII), powder
Sigma-Aldrich
Collagen from bovine achilles tendon, powder, suitable for substrate for collagenase
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I, solid, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I, (0.1% solution in 0.1 M acetic acid), aseptically processed, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from chicken sternal cartilage, Type II (Miller), powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen human, Bornstein and Traub Type I, acid soluble, powder, ~95% (SDS-PAGE)
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder
Supelco
Riboflavin (B2), analytical standard
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, solution, suitable for cell culture, High Performance
Riboflavin for peak identification, European Pharmacopoeia (EP) Reference Standard
Riboflavin, European Pharmacopoeia (EP) Reference Standard
USP
Riboflavin, United States Pharmacopeia (USP) Reference Standard
Supelco
Riboflavin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Collagen, Type I solution from rat tail, BioReagent, suitable for cell culture, sterile-filtered
Sigma-Aldrich
Collagen from rabbit skin, Bornstein and Traub Type I, powder
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type III (Sigma Type X), powder
Sigma-Aldrich
Collagen from Engelbreth-Holm-Swarm murine sarcoma basement membrane, Type IV (Miller), lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from bovine tracheal cartilage, Bornstein and Traub Type II, powder
Sigma-Aldrich
Collagen from bovine nasal septum, Bornstein and Traub Type II, powder
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type V (Sigma Type IX), powder
Sigma-Aldrich
Collagen from rat tail, Bornstein and Traub Type I, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
(−)-Riboflavin, meets USP testing specifications
Sigma-Aldrich
(−)-Riboflavin, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98%
Sigma-Aldrich
(−)-Riboflavin, from Eremothecium ashbyii, ≥98%