Merck
  • Delta-aminolevulinic acid dehydratase (ALAD) polymorphism in lead exposed Bangladeshi children and its effect on urinary aminolevulinic acid (ALA).
Delta-aminolevulinic acid dehydratase (ALAD) polymorphism in lead exposed Bangladeshi children and its effect on urinary aminolevulinic acid (ALA).
Environmental research (2014-12-03)
Saira Tasmin, Hana Furusawa, Sk Akhtar Ahmad, M H Faruquee, Chiho Watanabe
ABSTRACT
Lead has long been recognized as a harmful environmental pollutant. People in developing countries like Bangladesh still have a higher risk of lead exposure. Previous research has suggested that the delta-aminolevulinic acid dehydratase (ALAD) genotype can modify lead toxicity and individual susceptibility. As children are more susceptible to lead-induced toxicity, this study investigated whether the ALAD genotype influenced urinary excretion of delta-aminolevulinic acid (U-ALA) among children exposed to environmental lead in Bangladesh. Subjects were elementary schoolchildren from a semi-urban industrialized area in Bangladesh. A total of 222 children were studied. Blood and urine were collected to determine ALAD genotypes, blood lead levels and urinary aminolevulinic acid (U-ALA). The mean BPb level was 9.7 µg/dl for the study children. BPb was significantly positively correlated with hemoglobin (p<0.01). In total, allele frequency for ALAD 1 and 2 was 0.83 and 0.17 respectively. The mean U-ALA concentration was lower in ALAD1-2/2-2 carriers than ALAD1-1 carriers for boys (p=0.001). But for girls, U-ALA did not differ significantly by genotype (p=0.26). When U-ALA was compared by genotype at the same exposure level in a multiple linear regression analysis, boys who were ALAD1-2/2-2 carriers still had a lower level of U-ALA compared to ALAD1-1 carriers. This study provides information about the influence of ALAD polymorphism and its association with U-ALA in Bangladeshi children. Our results indicate that the ALAD1-2/2-2 genotype may have a protective effect in terms of U-ALA for environmentally lead exposed boys.
Sigma-Aldrich
Lead, powder, −325 mesh, ≥99% trace metals basis
Sigma-Aldrich
Creatinine, anhydrous, ≥98%
Lead, foil, not light tested, 50x50mm, thickness 0.004mm, 99.99+%
Supelco
Creatinine, Pharmaceutical Secondary Standard; Certified Reference Material
Lead, foil, light tested, 50x50mm, thickness 0.0125mm, 99.99+%
Lead, foil, light tested, 50x50mm, thickness 0.015mm, 99.99+%
Lead, foil, 4mm disks, thickness 0.025mm, 99.99+%
Lead, foil, 4mm disks, thickness 0.009mm, 99.99+%
Lead, foil, 4mm disks, thickness 0.02mm, 99.99+%
Lead, foil, 4mm disks, thickness 0.006mm, 99.99+%
Lead, foil, 25mm disks, thickness 1.0mm, as rolled, 99.99+%
Lead, foil, 2m coil, thickness 0.05mm, as rolled, 97% (sn 1.5%, sb 1.5%)
Lead, foil, 4mm disks, thickness 0.007mm, 99.99+%
Lead, foil, 2m coil, thickness 0.016mm, as rolled, 96% (sn 1.5%, sb 2.5%)
Lead, foil, 25mm disks, thickness 0.15mm, as rolled, 99.95%
Lead, foil, 25x25mm, thickness 1.0mm, as rolled, 99.99+%
Lead, foil, 8mm disks, thickness 0.007mm, 99.99+%
Lead, foil, 4mm disks, thickness 0.55mm, as rolled, 100%
Lead, foil, 10mm disks, thickness 0.01mm, 99.99+%
Lead, foil, 50x50mm, thickness 2.7mm, as rolled, 99.95%
Lead, foil, 1m coil, thickness 0.125mm, as rolled, 99.95%
Lead, foil, 8mm disks, thickness 0.05mm, 99.99+%
Lead, foil, 0.5m coil, thickness 0.125mm, as rolled, 99.95%
Lead, foil, 25mm disks, thickness 0.04mm, 99.99+%
Lead, foil, 1000x1000mm, thickness 0.5mm, as rolled, 99.95%
Lead, foil, 10mm disks, thickness 0.03mm, 99.99+%
Lead, foil, 10m coil, thickness 0.15mm, as rolled, 99.95%
Lead, foil, 10mm disks, thickness 0.006mm, 99.99+%
Lead, foil, 1m coil, thickness 0.5mm, as rolled, 99.95%
Lead, foil, 25mm disks, thickness 0.125mm, as rolled, 99.95%