Merck
  • Association between salivary cotinine and cardiovascular biomarkers among nonsmokers and current smokers: cross-sectional study of 10,081 participants.
Association between salivary cotinine and cardiovascular biomarkers among nonsmokers and current smokers: cross-sectional study of 10,081 participants.
European journal of vascular and endovascular surgery : the official journal of the European Society for Vascular Surgery (2014-12-03)
L Lu, D F Mackay, D E Newby, J P Pell
ABSTRACT
Both active smoking and exposure to secondhand smoke (SHS) are associated with cardiovascular disease, but sidestream smoke contains higher levels of small particles and toxic gases than mainstream smoke. The relationship between the concentration of cotinine and a number of cardiovascular biomarkers among nonsmokers and active smokers was examined. A cross-sectional study using the Scottish Health Surveys conducted between 1998 and 2010 was undertaken. Inclusion was restricted to participants aged ≥16 years who had provided saliva and blood samples. Uni- and multivariate regression models were used to examine the relationships between the concentration of cotinine and C-reactive protein (CRP), high-density lipoprotein (HDL) cholesterol, and fibrinogen concentrations, as well as total:HDL cholesterol ratios. Of the 10,018 eligible participants, 7,345 (73.3%) were confirmed to be nonsmokers (cotinine <15.0 ng/mL) and 2,673 (26.7%) were confirmed to be current smokers (cotinine ≥15.0 ng/mL). CRP and total:HDL cholesterol increased, and HDL cholesterol decreased, with increasing cotinine concentration across nonsmokers and smokers (all p < .001). However, there were step changes at the interface, whereby nonsmokers with a high exposure to SHS had lower concentrations of cotinine than light active smokers but comparable concentrations of CRP (p = .709), HDL cholesterol (p = .931), and total:HDL cholesterol (p = .405). Fibrinogen concentrations were significantly raised in moderate and heavy active smokers only (both p < .001). Exposure to SHS is associated with disproportionately higher biomarkers of cardiovascular risk compared with active smoking. Protection from exposure to SHS should be a public health priority.
Sigma-Aldrich
Fibrinogen from rat plasma, 60-80% protein (≥60% of protein is clottable)
Sigma-Aldrich
Fibrinogen from human plasma, 50-70% protein (≥80% of protein is clottable)
Sigma-Aldrich
Fibrinogen from human plasma, 35-65% protein (≥90% of protein is clottable).
Supelco
(−)-Cotinine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Cholesterol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Sigma-Aldrich
Cholesterol, from lanolin, ≥99.0% (GC)
Sigma-Aldrich
Cholesterol, tested according to Ph. Eur.
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
C Reactive Protein from human fluids, buffered aqueous solution
Sigma-Aldrich
(−)-Cotinine, ≥98%
Sigma-Aldrich
Fibrinogen from bovine plasma, Type I-S, 65-85% protein (≥75% of protein is clottable)
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications