Merck
  • Inhibition of bacterial mevalonate diphosphate decarboxylase by eriochrome compounds.
Inhibition of bacterial mevalonate diphosphate decarboxylase by eriochrome compounds.
Archives of biochemistry and biophysics (2014-12-17)
D Andrew Skaff, William J McWhorter, Brian V Geisbrecht, Gerald J Wyckoff, Henry M Miziorko
ABSTRACT
Mevalonate diphosphate decarboxylase (MDD; EC 4.1.1.33) catalyzes the irreversible decarboxylation of mevalonate diphosphate in the mevalonate pathway to form isopentenyl diphosphate, which is a precursor in the biosynthesis of many essential polyisoprenoid natural products, including sterols. In low G/C Gram-positive bacteria, which utilize the mevalonate pathway, MDD is required for cell viability and thus is a potential target for development of antibiotic drugs. To identify potential inhibitors of the enzyme, the National Cancer Institute's Mechanistic Diversity Set library of compounds was screened for inhibitors of Staphylococcus epidermidis MDD. From this screen, the compound Eriochrome Black A (EBA), an azo dye, was found to inhibit the enzyme with an IC50 value<5μM. Molecular docking of EBA into a crystal structure of S. epidermidis MDD suggested binding at the active site. EBA, along with the related Eriochrome B and T compounds, was evaluated for its ability to not only inhibit enzymatic activity but to inhibit bacterial growth as well. These compounds exhibited competitive inhibition towards the substrate mevalonate diphosphate, with Ki values ranging from 0.6 to 2.7μM. Non-competitive inhibition was observed versus ATP indicating binding of the inhibitor in the mevalonate diphosphate binding site, consistent with molecular docking predictions. Fluorescence quenching analyses also supported active site binding of EBA. These eriochrome compounds are effective at inhibiting S. epidermidis cell growth on both solid media and in liquid culture (MIC50 from 31 to 350μM) raising the possibility that they could be developed into antibiotic leads targeting pathogenic low-G/C Gram-positive cocci.
Sigma-Aldrich
(RS)-Mevalonic acid lithium salt, ≥96.0% (GC)
Supelco
(RS)-Mevalonic acid lithium salt, analytical standard
Sigma-Aldrich
Eriochrome® Black T, ACS reagent (indicator grade)