Merck
Sugarcoating Polycomb repression.
Developmental cell (2014-12-10)
C Peter Verrijzer
ABSTRACT
The addition of the monosaccharide O-GlcNAc is one of the most mysterious posttranslational modifications. In this issue of Developmental Cell, Gambetta and Müller (2014) show that O-GlcNAcylation of the Drosophila Polycomb group (PcG) protein Polyhomeotic (PH) is essential for homeotic gene silencing. O-GlcNAcylation counteracts nonproductive aggregation of PH, allowing transcriptional repression.
Product Description
Supelco
N-Acetylglucosamine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
N-Acetyl-D-glucosamine, ≥95% (HPLC)
Sigma-Aldrich
N-Acetyl-D-glucosamine, suitable for cell culture, BioReagent
Sigma-Aldrich
N-Acetyl-D-glucosamine, ≥99% (HPLC)