Merck
  • Extreme electric fields power catalysis in the active site of ketosteroid isomerase.
Extreme electric fields power catalysis in the active site of ketosteroid isomerase.
Science (New York, N.Y.) (2014-12-20)
Stephen D Fried, Sayan Bagchi, Steven G Boxer
ABSTRACT
Enzymes use protein architecture to impose specific electrostatic fields onto their bound substrates, but the magnitude and catalytic effect of these electric fields have proven difficult to quantify with standard experimental approaches. Using vibrational Stark effect spectroscopy, we found that the active site of the enzyme ketosteroid isomerase (KSI) exerts an extremely large electric field onto the C=O chemical bond that undergoes a charge rearrangement in KSI's rate-determining step. Moreover, we found that the magnitude of the electric field exerted by the active site strongly correlates with the enzyme's catalytic rate enhancement, enabling us to quantify the fraction of the catalytic effect that is electrostatic in origin. The measurements described here may help explain the role of electrostatics in many other enzymes and biomolecular systems.
Sigma-Aldrich
Activated Charcoal Norit®, Norit® CA1, wood, chemically activated, powder
Sigma-Aldrich
Activated Charcoal Norit®, Norit® SX ultra, from peat, steam activated and acid washed, highly purified, powder
Sigma-Aldrich
Activated Charcoal Norit®, Norit® PK 1-3, from peat, steam activated, granular
Sigma-Aldrich
Activated Charcoal Norit®, Norit® SX2, powder, from peat, multi-purpose activated charcoal, steam activated and acid washed
Sigma-Aldrich
Activated Charcoal Norit®, Norit® GAC 1240W, from coal, steam activated, granular
Sigma-Aldrich
Activated Charcoal Norit®, Norit® RB3, steam activated, rod
Carbon, rod, 150mm, diameter 10.0mm, graphite, 100%
Carbon, microleaf, 25x70mm, thinness 0.025μm, specific density 5μg/cm2, temporary glass support, 99.997%
Carbon, foil, 150x150mm, thickness 0.5mm, flexible graphite, 99.8%
Carbon, foil, 6mm disks, thickness 0.2mm, flexible graphite, 99.8%
Carbon, rod, 100mm, diameter 10.0mm, graphite, 100%
Carbon, rod, 300mm, diameter 3.0mm, graphite, 100%
Carbon, rod, 150mm, diameter 13.0mm, graphite, 100%
Carbon, microleaf, 25x70mm, thinness 0.050μm, specific density 10μg/cm2, temporary glass support, 99.997%
Carbon, foil, 5x10mm, thickness 2.0mm, hOpg
Carbon, tube, graphite, 100mm, outside diameter 3.18mm, inside diameter 1.18mm, wall thickness 1.0mm, 99.95%
Carbon, foil, 150x150mm, thickness 0.2mm, flexible graphite, 99.8%
Carbon, rod, 300mm, diameter 10.0mm, graphite, 100%
Carbon, foil, 500x500mm, thickness 0.2mm, flexible graphite, 99.8%
Carbon, foil, 500x500mm, thickness 0.5mm, flexible graphite, 99.8%
Carbon, foil, 8mm disks, thickness 0.2mm, flexible graphite, 99.8%
Carbon, rod, 150mm, diameter 2.0mm, graphite, 100%
Carbon, foil, 10x10mm, thickness 2.0mm, hOpg
Carbon, microleaf, 25x70mm, thinness 0.25μm, specific density 50μg/cm2, temporary glass support, 99.997%
Carbon, foil, 25x25mm, thickness 0.125mm, rigid graphite, fine grain size, 99.95%
Carbon, foil, 150x150mm, thickness 0.35mm, flexible graphite, 99.8%
Carbon, tube, graphite, 100mm, outside diameter 6.35mm, inside diameter 3.15mm, wall thickness 1.6mm, 99.95%
Carbon, rod, 2mm, diameter 0.5mm, graphite, 99.95%
Carbon, foil, 25mm disks, thickness 0.2mm, flexible graphite, 99.8%
Carbon, foil, 50x50mm, thickness 0.1mm, pyrolytic graphite, 99.99%