Merck
  • Master file size and apical transportation in severely curved root canals.
Master file size and apical transportation in severely curved root canals.
The New Zealand dental journal (2015-01-20)
S Moradi, R Disfani, H Baziar, F Daneshvar, H Jafarzadeh
ABSTRACT
A curvature leads to asymmetrical removal of root dentine, which results in an increase in the incidence of canal transportation. The aim of this study was to evaluate the effect of master apical file (MAF) size on the apical transportation (AT) of teeth with severe canal curvatures. Thirty-six mesial roots of human mandibular first molars were included. Inclusion criteria consisted of mature roots with closed apices, absence of carious lesions and resorptions, and root canal curvatures (CC) of 45 degrees < CC < 60 degrees. The root canals were prepared using a crown-down pressureless technique. The samples were equally divided into groups A, B, and C based on MAF size: group A: MAF size equivalent to #20; group B: MAF size equivalent to #25, and group C: MAF size equivalent to #30. Cone beam computed tomography was used to evaluate the AT. There were no statistically significant differences in the AT between the canals with different curvature angles up to MAF size #30 (P = 0.55). Using flexible hand files for canal preparation, an increase in MAF size up to #30 does not significantly influence AT in severely curved canals.
Product Description
USP
Edetate disodium, United States Pharmacopeia (USP) Reference Standard
Supelco
Ethylenediaminetetraacetic acid disodium salt dihydrate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, BioUltra, 98.5-101.5%
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, reagent grade, 98.5-101.5% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, ACS reagent, 99.0-101.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, suitable for electrophoresis, Molecular Biology, 99.0-101.0% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, Sigma Grade, suitable for plant cell culture, 98.5-101.5%
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, meets analytical specification of Ph. Eur., BP, USP, FCC, 99.0-101.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, ≥97.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid calcium disodium salt hydrate
Sigma-Aldrich
Sodium hypochlorite solution, 6-14% active chlorine basis
Sigma-Aldrich
Sodium hypochlorite solution, purum, ~10% (RT)
Sigma-Aldrich
Sodium hypochlorite solution, reagent grade, available chlorine 10-15 %
Disodium edetate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, Molecular Biology, 0.5 M in H2O, DNase, RNase, NICKase and protease, none detected
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, 2%, solution
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.4-100.6%, powder
Sodium calcium edetate, European Pharmacopoeia (EP) Reference Standard