Merck
  • hARIP2 is a putative growth-promoting factor involved in human colon tumorigenesis.
hARIP2 is a putative growth-promoting factor involved in human colon tumorigenesis.
Asian Pacific journal of cancer prevention : APJCP (2014-11-07)
Rui-Feng Gao, Zhan-Dong Li, Jing Jiang, Li-Hua Yang, Ke-Tong Zhu, Rui-Xin Lin, Hao Li, Quan Zhao, Nai-Sheng Zhang
ABSTRACT
Activin is a multifunctional growth and differentiation factor of the growth factor-beta (TGF-β) superfamily, which inhibits the proliferation of colon cancer cells. It induces phosphorylation of intracellular signaling molecules (Smads) by interacting with its type I and type II receptors. Previous studies showed that human activin receptor-interacting protein 2 (hARIP2) can reduce activin signaling by interacting with activin type II receptors; however, the activity of hARIP2 in colon cancer has yet to be detailed. In vitro, overexpression of hARIP2 reduced activin-induced transcriptional activity and enhanced cell proliferation and colony formation in human colon cancer HCT8 cells and SW620 cells. Also, hARIP2 promoted colon cancer cell apoptosis, suggesting that a vital role in the initial stage of colon carcinogenesis. In vivo, immunohistochemistry revealed that hARIP2 was expressed more frequently and much more intensely in malignant colon tissues than in controls. These results indicate that hARIP2 is involved in human colon tumorigenesis and could be a predictive maker for colon carcinoma aggressiveness.
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
Ethanol, purum, absolute ethanol, denaturated with 2% 2-butanone, A15 MEK1, ≥99.8% (based on denaturant-free substance)
Supelco
Ethanol, standard for GC
Sigma-Aldrich
Ethanol, tested according to Ph. Eur.
Sigma-Aldrich
Ethanol, purum, absolute ethanol, denaturated with 4.8% isopropanol, A15 IPA1, ≥99.8% (based on denaturant-free substance)
Sigma-Aldrich
Ethanol, BioUltra, Molecular Biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethanol
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)