Merck
  • Knockdown of YAP1 inhibits the proliferation of osteosarcoma cells in vitro and in vivo.
Knockdown of YAP1 inhibits the proliferation of osteosarcoma cells in vitro and in vivo.
Oncology reports (2014-07-06)
Zhi Yang, Ming Zhang, Ke Xu, Lin Liu, Wei-Kun Hou, Yuan-Zhen Cai, Peng Xu, Jian-Feng Yao
ABSTRACT
Yes-associated protein 1 (YAP1) is a candidate oncogene that is involved in tumorigenesis and progression of many malignant tumors. Recently, many studies have revealed that YAP1 is highly expressed in human osteosarcoma. To investigate the role of YAP1 in osteosarcoma tumorigenesis, the expression of YAP1 in the osteosarcoma cell lines (MG-63 and HOS) was knocked down by small hairpin RNA (shRNA), and the cell proliferation and colony formation assay showed that knockdown of YAP1 significantly suppressed the cell proliferation and colony formation of osteosarcoma cells. Subsequently, cell cycle distribution was analyzed by flow cytometry, and the results showed an accumulation of YAP1-knockdown cells in the G0/G1 phase, suggesting that YAP1 knockdown results in the arrest of cell cycle progression. Additionally, the knockdown of YAP1 also inhibited the tumorsphere formation in vitro and the growth of xenograft tumors in vivo. Therefore, these data suggest that YAP1 knockdown inhibits the proliferation of osteosarcoma cells. However, the mechanism of action was unclear. Further investigation showed that in the YAP1-knockdown MG-63 and HOS cells, the level of cylinD1 and c-myc expression, target genes of the Wnt signaling pathway and TOP-Flash reporter activity were all significantly decreased, which indicated that the inhibitory effect of YAP1 knockdown on osteosarcoma might be associated with the Wnt signaling pathway. Taken together, our results demonstrated that YAP1 is an important regulator of osteosarcoma tumorigenesis and knockdown of YAP1 would be a novel therapeutic strategy for osteosarcoma.
Product Description
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)