Merck
  • Evaluation of the pharmacokinetics of the DPP-4 inhibitor gemigliptin when coadministered with rosuvastatin or irbesartan to healthy subjects.
Evaluation of the pharmacokinetics of the DPP-4 inhibitor gemigliptin when coadministered with rosuvastatin or irbesartan to healthy subjects.
Current medical research and opinion (2014-10-29)
Hee Youn Choi, Hyeong-Seok Lim, Yo Han Kim, Hae Sun Jeon, Mi Jo Kim, Shi Hyang Lee, Jong Hyuk Jung, Young Kyoung Lee, Hyun Jeong Kim, Kyun-Seop Bae
ABSTRACT
Gemigliptin is a selective DPP4 inhibitor used to treat type 2 diabetes. The objective of this study was to evaluate the pharmacokinetics (PKs) of gemigliptin, rosuvastatin, and irbesartan monotherapies and combination therapies. Randomized, open-label, three-treatment, six-sequence, three-period, crossover studies were performed on healthy male volunteers. The three treatments were: 50 mg gemigliptin alone; 20 mg rosuvastatin (part A) or 300 mg irbesartan alone (part B); and rosuvastatin or irbesartan with concomitant gemigliptin. Each drug was administered as part of once daily, 7 day, repeated dosing regimens with a 14 day washout period. NCT01823133 (part A) and NCT01825850 (part B). The primary PK parameters - Cmax and AUCτ - were compared to the geometric mean ratios (GMRs) and 90% confidence intervals (90% CIs) that were determined for the combination therapies and monotherapies. A total of 60 participants were administered the study drugs, and 52 participants (27 participants in part A; 25 participants in part B) were analyzed as part of the PK dataset. In part A, the GMRs (gemigliptin + rosuvastatin/gemigliptin) of the Cmax and AUCτ values of gemigliptin were 0.955 (90% CI = 0.874-1.044) and 1.023 (90% CI = 0.991-1.057), and those of rosuvastatin were 1.012 (90% CI = 0.946-1.084) and 1.086 (90% CI = 1.032-1.142), respectively. In part B, the GMRs of the Cmax and AUCτ values of gemigliptin were 1.046 (90% CI = 0.964-1.134) and 1.035 (90% CI = 1.005-1.065), and those of irbesartan were 0.966 (90% CI = 0.897-1.040) and 1.050 (90% CI = 0.993-1.111), respectively. The limitations of this study include its relatively short treatment period and small sample size, as only healthy participants were included. Gemigliptin does not affect the PK properties of rosuvastatin or irbesartan; also, rosuvastatin and irbesartan do not affect the PKs of gemigliptin.
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%