Merck
  • Use of non-steroidal anti-inflammatory drugs and risk of chronic kidney disease in people with Type 2 diabetes mellitus, a nationwide longitudinal cohort study.
Use of non-steroidal anti-inflammatory drugs and risk of chronic kidney disease in people with Type 2 diabetes mellitus, a nationwide longitudinal cohort study.
Diabetic medicine : a journal of the British Diabetic Association (2014-10-15)
H-J Tsai, Y-H Hsu, Y-W Huang, Y-K Chang, J-S Liu, C-C Hsu
ABSTRACT
To investigate the temporal relationship between non-steroidal anti-inflammatory drug use and the development of chronic kidney disease in people with Type 2 diabetes mellitus. We conducted a retrospective cohort study and followed up a population with Type 2 diabetes who were chronic kidney disease-free (n = 48,715) using national health insurance claims data in Taiwan. Exposure status to non-steroidal anti-inflammatory drugs in 2007 was measured. A total of 6406 subjects with incident chronic kidney disease were identified from the period 2008 to 2011. Multivariable proportional hazards models were applied to determine the temporal relationship between non-steroidal anti-inflammatory drug use and the development of chronic kidney disease. We observed a significant temporal relationship between non-steroidal anti-inflammatory drug use and the development of chronic kidney disease in people with Type 2 diabetes. Compared with people not taking any non-steroidal anti-inflammatory drug in 2007, those who were taking such drugs for at least 90 days in 2007 had a higher risk of chronic kidney disease development (adjusted hazard ratio 1.37, 95% CI 1.26-1.49). In subgroup analyses, those people (irrespective of age, sex, various comorbidities and use of anti-hypertensive drugs, aspirin or acetaminophen) who were taking non-steroidal anti-inflammatory drugs for at least 90 days were more likely to develop chronic kidney disease than people who were not taking any non-steroidal anti-inflammatory drug. The results suggest that there is a positive temporal relationship between non-steroidal anti-inflammatory drug use and increased risk of chronic kidney disease in people with Type 2 diabetes. The use of non-steroidal anti-inflammatory drugs should be based on clinical evaluations of benefits and risks, and should be prescribed with caution for people with Type 2 diabetes.
Product Description
Supelco
Acetaminophen, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetaminophen, BioXtra, ≥99.0%
USP
Acetaminophen, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetaminophen, analytical standard
Sigma-Aldrich
Acetaminophen, meets USP testing specifications, 98.0-102.0%, powder
Supelco
Acetaminophen solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Paracetamol, European Pharmacopoeia (EP) Reference Standard