Merck
  • Glycosylation and subsequent malonylation of isoflavonoids in E. coli: strain development, production and insights into future metabolic perspectives.
Glycosylation and subsequent malonylation of isoflavonoids in E. coli: strain development, production and insights into future metabolic perspectives.
Journal of industrial microbiology & biotechnology (2014-09-06)
Niranjan Koirala, Ramesh Prasad Pandey, Duong Van Thang, Hye Jin Jung, Jae Kyung Sohng
ABSTRACT
Genistin and daidzein exhibit a protective effect on DNA damage and inhibit cell proliferation. Glycosylation and malonylation of the compounds increase water solubility and stability. Constructed pET15b-GmIF7GT and pET28a-GmIF7MAT were used for the transformation of Escherichia coli and bioconversion of genistein and daidzein. To increase the availability of malonyl-CoA, a critical precursor of GmIF7MAT, genes for the acyl-CoA carboxylase α and β subunits (nfa9890 and nfa9940), biotin ligase (nfa9950), and acetyl-CoA synthetase (nfa3550) from Nocardia farcinia were also introduced. Thus, the isoflavonoids were glycosylated at position 7 by 7-O-glycosyltranferase and were further malonylated at position 6(″) of glucose by malonyl-CoA: isoflavone 7-O-glucoside-6(″)-O-malonyltransferase both from Glycine max. Engineered E. coli produced 175.7 µM (75.90 mg/L) of genistin and 14.2 µM (7.37 mg/L) genistin 6''-O-malonate. Similar conditions produced 162.2 µM (67.65 mg/L) daidzin and 12.4 µM (6.23 mg/L) daidzin 6''-O-malonate when 200 µM of each substrate was supplemented in the culture. Based on our findings, we speculate that isoflavonoids and their glycosides may prove useful as anticancer drugs with added advantage of increased solubility, stability and bioavailability.
Product Description
Supelco
Genistein, analytical standard
Sigma-Aldrich
Genistein, from Glycine max (soybean), ~98% (HPLC)
Sigma-Aldrich
Genistein, synthetic, ≥98% (HPLC), powder
Sigma-Aldrich
Formononetin, ≥99.0% (TLC)
Supelco
Daidzein, analytical standard
Sigma-Aldrich
Daidzein, ≥98%, synthetic