Merck
  • Occurrence of contaminants of emerging concern along the California coast (2009-10) using passive sampling devices.
Occurrence of contaminants of emerging concern along the California coast (2009-10) using passive sampling devices.
Marine pollution bulletin (2013-05-21)
David A Alvarez, Keith A Maruya, Nathan G Dodder, Wenjian Lao, Edward T Furlong, Kelly L Smalling
ABSTRACT
Three passive sampling devices (PSDs), polar organic chemical integrative samplers (POCIS), polyethylene devices (PEDs), and solid-phase microextraction (SPME) samplers were used to sample a diverse set of chemicals in the coastal waters of San Francisco Bay and the Southern California Bight. Seventy one chemicals (including fragrances, phosphate flame retardants, pharmaceuticals, PAHs, PCBs, PBDEs, and pesticides) were measured in at least 50% of the sites. The chemical profile from the San Francisco Bay sites was distinct from profiles from the sites in the Southern California Bight. This distinction was not due to a single compound or class, but by the relative abundances/concentrations of the chemicals. Comparing the PSDs to mussel (Mytilus spp.) tissues, a positive correlation exists for the 25 and 26 chemicals in common for the PEDs and SPME, respectively. Diphenhydramine was the only common chemical out of 40 analyzed in both POCIS and tissues detected at a common site.
Sigma-Aldrich
Sodium sulfate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium sulfate, BioUltra, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Sodium sulfate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium sulfate, ACS reagent, ≥99.0%, anhydrous, granular
Sigma-Aldrich
Sodium sulfate, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Sodium sulfate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium sulfate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Sodium sulfate, ACS reagent, ≥99.0%, anhydrous, powder
Sigma-Aldrich
Sodium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Hexamethyldisiloxane, viscosity 0.65 cSt (25 °C)
Sigma-Aldrich
Poly(dimethylsiloxane), viscosity 1.0 cSt (25 °C)
Sigma-Aldrich
Sodium sulfate, puriss., meets analytical specification of Ph. Eur., BP, USP, anhydrous, 99.0-100.5% (calc. to the dried substance)
Sigma-Aldrich
Sodium sulfate, ≥99.0%, suitable for plant cell culture
Sigma-Aldrich
Hexane, ReagentPlus®, ≥99%
Sigma-Aldrich
Hexane, suitable for HPLC, ≥95%
Sigma-Aldrich
Hexane, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99% (GC)
Supelco
Hexane, analytical standard
Sigma-Aldrich
Hexane, anhydrous, 95%
Sigma-Aldrich
Hexane, suitable for HPLC, ≥97.0% (GC)
Sigma-Aldrich
Hexane, HPLC Plus, for HPLC, GC, and residue analysis, ≥95%
Sigma-Aldrich
Hexane, Laboratory Reagent, ≥95%