Merck
  • Toxicity induced by F. poae-contaminated feed and the protective effect of Montmorillonite supplementation in broilers.
Toxicity induced by F. poae-contaminated feed and the protective effect of Montmorillonite supplementation in broilers.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2014-10-09)
Lingchen Yang, Zhiyong Zhao, Yifeng Deng, Zhenlei Zhou, Jiafa Hou
ABSTRACT
The T-2 and HT-2 toxins, the main metabolites of Fusarium poae, induce toxicity in broilers and accumulate in tissues. Consequently, during the breeding process of broilers, diets are frequently supplemented with physical adsorbents to protect birds against the toxicity induced by mycotoxins. In the present research, T-2 and HT-2 were produced in maize inoculated with F. poae. Mont, the strongest adsorbent based on in vitro adsorption ratios, was added to the contaminated diet. One-day-old chickens were randomly and equally divided into the following four groups: control diet group, Mont supplemented diet group, contaminated diet group and detoxification diet group. The experiment lasted for 42 days. Compared to the control group, the contaminated group showed significant decrease in body weight, feed intake and TP (P < 0.05), and marked increase in FCR, ALP, AST and ALT activity, T-2/HT-2 residues in the tissues and the relative expressions of apoptosis-related mRNAs (P < 0.05). Mont supplementation provided protection for the treated broilers in terms of performance, blood biochemistry, hepatic function, T-2/HT-2 residue of tissues and apoptosis. Therefore, Mont may be suitable as a detoxification agent for T-2/HT-2 in feed for broilers.
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Supelco
Mycophenolic acid, analytical standard
Sigma-Aldrich
Beauvericin, ≥97% (HPLC)
Sigma-Aldrich
Mycophenolic acid, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Deoxynivalenol
Sigma-Aldrich
Aflatoxin G2
Sigma-Aldrich
Mycophenolic acid, ≥98%
Sigma-Aldrich
Ethyl acetate, anhydrous, 99.8%
Sigma-Aldrich
Ethyl acetate, natural, ≥99%, FCC, FG
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Nivalenol in acetonitrile, IRMM®, certified reference material
Supelco
Diacetoxyscirpenol solution, ~100 μg/mL in acetonitrile, analytical standard
Sigma-Aldrich
Ethyl acetate, ReagentPlus®, ≥99.8%
Sigma-Aldrich
Ethyl acetate, ≥99%, FCC, FG
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Ethyl acetate, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Ethyl acetate, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
Ethyl acetate, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Acetonitrile, suitable for HPLC-GC, ≥99.8% (GC)
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Supelco
Ethyl Acetate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethyl acetate
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis