Merck
  • Relationship between serum levels of tau fragments and clinical progression of Alzheimer's disease.
Relationship between serum levels of tau fragments and clinical progression of Alzheimer's disease.
Journal of Alzheimer's disease : JAD (2014-08-30)
Kim Henriksen, Inger Byrjalsen, Claus Christiansen, Morten Asser Karsdal
ABSTRACT
Enzyme-generated fragments of tau have been linked to neuronal death and may serve as serum biomarkers of cognitive loss. Two competitive ELISAs detecting an ADAM10-generated fragment (Tau-A) or a caspase-3-generated fragment (Tau-C) were measured in baseline serum samples from patients with mild to moderate Alzheimer's disease (AD) from a Phase III clinical trial, and correlated to change in the Alzheimer's Disease Assessment Scale-Cognitive subscale (ADAS-Cog11) and Clinical Dementia Rating-Sum of Boxes (CDR-SB) over a 64-week period using an MMRM-analysis. Relationship between the biomarkers and changes in ADAS-Cog11 score as a function of time were observed for Tau-C and change in ADAS-Cog11 (p = 0.06), and for Tau-A and change in CDR-SB (p = 0.04). The correlation of Tau-A/Tau-C ratio with cognitive change assessed by ADAS-Cog11 was even more significant (p < 0.006). These data indicate that measuring the balance between tau fragments in serum may provide a marker of the rate of progression of AD and warrant studies in larger cohorts.
Product Description
Supelco
Sulfuric acid concentrate, 0.1 M H2SO4 in water (0.2N), eluent concentrate for IC
Supelco
Sulfuric acid, for the determination of nitrogen, ≥97.0%
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sulfuric acid, 99.999%
Sigma-Aldrich
Sulfuric acid, ACS reagent, 95.0-98.0%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Sulfuric acid, puriss., meets analytical specification of Ph. Eur., BP, 95-97%
Sigma-Aldrich
Sulfuric acid, puriss. p.a., for determination of Hg, ACS reagent, reag. ISO, reag. Ph. Eur., 95.0-97.0%
Sigma-Aldrich
Sulfuric acid solution, puriss. p.a., ≥25% (T)
Sigma-Aldrich
Sulfuric acid, puriss., meets analytical specification of Ph. Eur., BP, 95-97%