Merck
  • Lignan and flavonoids from the stems of Zea mays and their anti-inflammatory and neuroprotective activities.
Lignan and flavonoids from the stems of Zea mays and their anti-inflammatory and neuroprotective activities.
Archives of pharmacal research (2014-04-22)
Ye-Jin Jung, Ji-Hae Park, Jin-Gyeong Cho, Kyeong-Hwa Seo, Dong-Sung Lee, Youn-Chul Kim, Hee-Cheol Kang, Myoung-Chong Song, Nam-In Baek
ABSTRACT
The stems of Zea mays L., otherwise known as cornstalks, were extracted with 80 % aqueous MeOH, and the concentrated extract was successively partitioned with ethyl acetate (EtOAc), normal butanol, and water. From the EtOAc fraction, a new lignan along with three known flavonoids, tricin (1), salcolin A (2), and salcolin B (3), were isolated. The chemical structure of the lignan was determined to be tetrahydro-4,6-bis(4-hydroxy-3-methoxyphenyl)-1H,3H-furo[3,4-c]furan-1-one (4) through spectroscopic data analyses including NMR, MS, and IR. All compounds were isolated for the first time from this plant. The isolated compounds were evaluated for their inhibitory activity against NO production in Lipopolysaccharide-induced RAW 264.7 cells and their protective activity in glutamate-induced cell death in HT22 cells. The compounds 1, 2 and 4 showed anti-inflammatory effects with IC50 values of 2.63, 14.65, and 18.91 μM, respectively, as well as neuroprotective effects with EC50 values of 25.14, 47.44, and >80 μM, respectively.
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Supelco
Hydrogen chloride – ethanol, ~1.25 M HCl, derivatization grade (GC derivatization), LiChropur
Supelco
Hydrogen chloride – methanol solution, ~1.25 m HCl (T), derivatization grade (GC derivatization), LiChropur
Supelco
Hydrogen chloride – 2-propanol solution, ~1.25 M HCl (T), derivatization grade (GC derivatization), LiChropur
Sigma-Aldrich
Hydrogen chloride solution, 4.0 M in dioxane
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Hydrochloric acid solution, for amino acid analysis, ~6 M in H2O
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in acetic acid
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
Sulfuric acid solution, puriss. p.a., ≥25% (T)
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrochloric acid, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
Hydrochloric acid, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Sulfuric acid, ACS reagent, 95.0-98.0%
Sigma-Aldrich
Hydrochloric acid, puriss., 24.5-26.0%
Sigma-Aldrich
Hydrochloric acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., fuming, ≥37%, APHA: ≤10
Sigma-Aldrich
Sulfuric acid, puriss., meets analytical specification of Ph. Eur., BP, 95-97%
Sigma-Aldrich
Sulfuric acid, puriss. p.a., for determination of Hg, ACS reagent, reag. ISO, reag. Ph. Eur., 95.0-97.0%
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Sulfuric acid, puriss., meets analytical specification of Ph. Eur., BP, 95-97%
Supelco
Sulfuric acid, for the determination of nitrogen, ≥97.0%
Sigma-Aldrich
Sulfuric acid, 99.999%
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Hydrogen chloride solution, 2.0 M in diethyl ether
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in diethyl ether
Sigma-Aldrich
2-Propanol, Laboratory Reagent, ≥99.5%