Merck
  • Exacerbation of intracranial aneurysm and aortic dissection in hypertensive rat treated with the prostaglandin F-receptor antagonist AS604872.
Exacerbation of intracranial aneurysm and aortic dissection in hypertensive rat treated with the prostaglandin F-receptor antagonist AS604872.
Journal of pharmacological sciences (2014-10-25)
Miyuki Fukuda, Tomohiro Aoki, Toshiaki Manabe, Akiko Maekawa, Takayuki Shirakawa, Hiroharu Kataoka, Yasushi Takagi, Susumu Miyamoto, Shuh Narumiya
ABSTRACT
Intracranial aneurysm (IA) and aortic dissection are both complications of hypertension and characterized by degeneration of the media. Given the involvement of prostaglandin F2α and its receptor, FP, in extracellular matrix remodeling in a mouse model of pulmonary fibrosis, here we induced hypertension and IA in rats by salt loading and hemi-lateral ligation of renal and carotid arteries and examined effects of a selective FP antagonist, AS604872, on these vascular events. AS604872 significantly accelerated degeneration of the media in both cerebral artery and aorta as evidenced by thinning of the media and disruption of the elastic lamina and promoted IA and aortic dissection. Notably, AS604872 induced expression of pro-inflammatory genes such as E-selectin in lesions and significantly enhanced macrophage infiltration. Suppression of surface expression of E-selectin with cimetidine prevented macrophage infiltration and aortic dissection. Thus, AS604872 exacerbates vascular inflammation in hypertensive rats and facilitates IA and aortic dissection. These results demonstrate that both IA and aortic dissection are caused by chronic inflammation of the arterial wall, which is worsened by AS604872, cautioning that other FP antagonists may share such deleterious actions in vascular homeostasis and suggesting that AS604872 can be used to make models of these vascular diseases with extensive degeneration.
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Supelco
Cimetidine, Pharmaceutical Secondary Standard; Certified Reference Material
Cimetidine, European Pharmacopoeia (EP) Reference Standard
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Cimetidine, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Cimetidine
Sigma-Aldrich
Anti-AKR1C3 antibody, Mouse monoclonal, clone NP6.G6.A6, purified from hybridoma cell culture
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Fluorescein, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Fluorescein, free acid
Cimetidine for peak identification, European Pharmacopoeia (EP) Reference Standard
Cimetidine for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
3-amino-propionitrile, AldrichCPR