Merck
  • Selective pressurized liquid extraction technique for halogenated organic pollutants in marine mammal blubber: a lipid-rich matrix.
Selective pressurized liquid extraction technique for halogenated organic pollutants in marine mammal blubber: a lipid-rich matrix.
Journal of chromatography. A (2015-02-14)
E M Robinson, M Jia, S J Trumble, S Usenko
ABSTRACT
Analytical methods for unique and rare samples, such as marine mammal tissue, strive to reduce opportunities for analyte loss and contamination. Historically, analytical methodologies for marine mammal tissues required an extraction followed by multiple cleanup and concentration steps. These steps increase the opportunity for analyte loss and sample contamination. Selective pressurized liquid extractions (SPLE; an analytical technique that combines PLE with in-cell adsorbent cleanup) have the potential to reduce and/or eliminate the number of steps. A SPLE method was developed for the simultaneous extraction of polybrominated diphenyl ethers (PBDEs), polychlorinated biphenyls (PCBs), and organochlorine pesticides (OCPs) from bowhead whale blubber. This SPLE utilized acidic silica with a fat-to-fat retainer ratio of 0.02 as well as eliminated post-extraction cleanup steps, such as size-exclusion chromatography step. In addition, neutral silica was placed beneath the acidic silica as an acid buffer, thereby preventing acid from contaminating the extraction system. Analysis was performed using gas chromatography/mass spectrometry in electron capture negative ionization mode. PBDE, PCB and OCP triplicate recoveries averaged 84±1%, 83±3%, and 76±11%, respectively. Overall, measurements of NIST Whale Blubber SRM 1945 were within±30% of certified values. PBDEs were measured for the first time in bowhead whale blubber; average concentrations ranged from 0.2 to 1.4 ng g(-1) wet weight (ww). Average OCPs and PCBs concentrations ranged from 0.4 to 37 ng g(-1)ww and 0.1 to 3.0 ng g(-1)ww, respectively, which were within one order of magnitude lower than those previously reported in bowhead whale blubber.
Product Description
Supelco
Sulfuric acid concentrate, 0.1 M H2SO4 in water (0.2N), eluent concentrate for IC
Supelco
Sulfuric acid, for the determination of nitrogen, ≥97.0%
Sigma-Aldrich
Sulfuric acid, 99.999%
Sigma-Aldrich
Sulfuric acid, ACS reagent, 95.0-98.0%
Sigma-Aldrich
Sulfuric acid, puriss., meets analytical specification of Ph. Eur., BP, 95-97%
Sigma-Aldrich
Sulfuric acid, puriss. p.a., for determination of Hg, ACS reagent, reag. ISO, reag. Ph. Eur., 95.0-97.0%
Sigma-Aldrich
Sulfuric acid solution, puriss. p.a., ≥25% (T)
Sigma-Aldrich
Sulfuric acid, puriss., meets analytical specification of Ph. Eur., BP, 95-97%