Merck
Pharmacokinetics of raltegravir in the semen of HIV-infected men.
Antiviral therapy (2014-02-13)
Tony Antoniou, Mona R Loutfy, Jason Brunetta, Graham Smith, Roberta Halpenny, Charles la Porte
ABSTRACT
We sought to determine the pharmacokinetic disposition of raltegravir in the blood and seminal plasma of HIV-infected men. We conducted a pharmacokinetic study using a staggered sampling approach. A total of 16 HIV-infected men receiving raltegravir-based therapy were recruited into the study. Each participant provided six blood plasma and six seminal plasma samples for quantification of drug concentrations in both compartments. Blood and semen samples were obtained within 1 h of each other and were collected prior to the morning dose and at 1, 2, 4, 8 and 12 h post-ingestion. Drug concentrations were determined by liquid chromatography tandem mass spectrometry. A total of 96 semen samples and 96 blood samples were obtained from all participants during the study period. The median age and baseline CD4(+) T-cell count of the study participants were 48 years (IQR 42-53) and 450 cells/mm(3) (IQR 289-585). Virological suppression to <50 copies/ml had been maintained for a median of 21 months (IQR 7-35) at the time of study enrolment. The median seminal plasma-to-blood plasma ratios and AUC0-12 h seminal plasma-to-blood plasma ratios of raltegravir were 3.25 (IQR 1.46-5.37) and 2.26 (IQR 1.05-4.45), respectively. Concentrations of raltegravir in seminal plasma are several fold higher than those attained in blood plasma and those required to inhibit viral replication in this compartment. Further research examining the therapeutic and prophylactic implications of these findings is warranted.
Product Description
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Quinoxaline, ≥98%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, suitable for HPLC-GC, ≥99.8% (GC)
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Ultrapure Acetonitrile
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, ≥99.8% (GC)
Sigma-Aldrich
Acetonitrile, ≥99.5% (GC)
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard