Merck
  • Engraftable neural crest stem cells derived from cynomolgus monkey embryonic stem cells.
Engraftable neural crest stem cells derived from cynomolgus monkey embryonic stem cells.
Biomaterials (2014-12-06)
Weiqiang Li, Lihua Huang, Wanyi Lin, Qiong Ke, Rui Chen, Xingqiang Lai, Xiaoyu Wang, Jifeng Zhang, Meihua Jiang, Weijun Huang, Tao Wang, Xuesong Yang, Yuan Chen, Wu Song, Andy Peng Xiang
ABSTRACT
Neural crest stem cells (NCSCs), a population of multipotent cells that migrate extensively and give rise to diverse derivatives, including peripheral and enteric neurons and glia, craniofacial cartilage and bone, melanocytes and smooth muscle, have great potential for regenerative medicine. Non-human primates provide optimal models for the development of stem cell therapies. Here, we describe the first derivation of NCSCs from cynomolgus monkey embryonic stem cells (CmESCs) at the neural rosette stage. CmESC-derived neurospheres replated on polyornithine/laminin-coated dishes migrated onto the substrate and showed characteristic expression of NCSC markers, including Sox10, AP2α, Slug, Nestin, p75, and HNK1. CmNCSCs were capable of propagating in an undifferentiated state in vitro as adherent or suspension cultures, and could be subsequently induced to differentiate towards peripheral nervous system lineages (peripheral sympathetic neurons, sensory neurons, and Schwann cells) and mesenchymal lineages (osteoblasts, adipocytes, chondrocytes, and smooth muscle cells). CmNCSCs transplanted into developing chick embryos or fetal brains of cynomolgus macaques survived, migrated, and differentiated into progeny consistent with a neural crest identity. Our studies demonstrate that CmNCSCs offer a new tool for investigating neural crest development and neural crest-associated human disease and suggest that this non-human primate model may facilitate tissue engineering and regenerative medicine efforts.
Product Description
Supelco
Calcium ion solution for ISE, 0.1 M Ca, analytical standard (for ion-selective electrodes)
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
USP
Ascorbic acid, United States Pharmacopeia (USP) Reference Standard
Supelco
Ascorbic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
Calcium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Calcium chloride
Sigma-Aldrich
Calcium chloride, powder, 99.99% trace metals basis
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Magnesium chloride, suitable for insect cell culture, BioReagent, ≥97.0%
Sigma-Aldrich
Magnesium chloride, ≥98%
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Supelco
Dexamethasone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Dexamethasone, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Supelco
Dexamethasone, VETRANAL®, analytical standard
Sigma-Aldrich
Dexamethasone, meets USP testing specifications
Ascorbic acid, European Pharmacopoeia (EP) Reference Standard
Dexamethasone, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder
USP
Dexamethasone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dexamethasone, tested according to Ph. Eur.
Dexamethasone for system suitability, European Pharmacopoeia (EP) Reference Standard
Dexamethasone, British Pharmacopoeia (BP) Assay Standard
Supelco
4-tert-Octylphenol monoethoxylate solution, 10 μg/mL in acetone, analytical standard
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Supelco
L-Ascorbic acid, analytical standard