Merck
  • Identification of the bioactive components of orally administered Lithocarpus polystachyus Rehd and their metabolites in rats by liquid chromatography coupled to LTQ Orbitrap mass spectrometry.
Identification of the bioactive components of orally administered Lithocarpus polystachyus Rehd and their metabolites in rats by liquid chromatography coupled to LTQ Orbitrap mass spectrometry.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2014-06-03)
Xiong Li, Ya Zhao, Shaozhen Hou, Song Huang, Weiqin Yang, Xiaoping Lai, Xing Zeng
ABSTRACT
A rapid, sensitive and selective UPLC/MS method using LTQ Orbitrap mass spectrometry was established for the analysis and characterization of the main biological components and their metabolites in rat plasma, urine and feces following oral administration of Lithocarpus polystachyus extract. In vivo, 22 flavonoid metabolites were observed in rat plasma, and 13 metabolites were detected in rat urine, whereas just two aglycones of dihydrochalcone (3-hydroxy phloretin and phloretin) could be detected in rat feces. Among these metabolites, one new and a known dihydrochalcone metabolite were isolated and definitely identified. Besides, five dihydrochalcone metabolites were tentatively identified as new compounds. A metabolism study of 3-hydroxyphlorizin and phloridzin was also conducted. Glucuronidation was the main metabolic pathway of phloridzin, whereas glucuronidation and sulfonation were the main metabolic pathway of 3-hydroxyphlorizin. These results provided a basis for evaluating the bioactive components of a complex natural medicine and their mechanisms of actions.
Product Description
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard