Merck
  • The influence of AAV2-mediated gene transfer of human IL-10 on neurodegeneration and immune response in a murine model of Parkinson's disease.
The influence of AAV2-mediated gene transfer of human IL-10 on neurodegeneration and immune response in a murine model of Parkinson's disease.
Pharmacological reports : PR (2014-06-21)
Ilona Joniec-Maciejak, Agnieszka Ciesielska, Adriana Wawer, Anna Sznejder-Pachołek, Joanna Schwenkgrub, Agnieszka Cudna, Piotr Hadaczek, Kristof S Bankiewicz, Anna Członkowska, Andrzej Członkowski
ABSTRACT
The aim of this study was to examine the effect of AAV2-hIL-10 (vector containing cDNA for human interleukin 10) on dopaminergic system activity (measured as DA levels and TH mRNA expression in mouse striata), and other monoamine and amino acid neurotransmitters concentration as well as development of inflammatory processes (measured as TGF-β, IFN-γ and GFAP mRNA expression) in a murine MPTP neurotoxicant model of Parkinson's disease. Male C57BL/6 mice 12 months-old were used in this study. AAV2-hIL-10 vector was bilaterally administered into striatum at 14, 21 or 28 days prior to MPTP intoxication. Animals were sacrificed at 7 days following MPTP injection. The expression of hIL-10 (human interleukin 10) was examined by ELISA. Striatal monoamine and amino acid neurotransmitters were measured by HPLC method. TH, TGF-β, IFN-γ and GFAP mRNA expression was examined by RT-PCR method. MPTP treatment dramatically reduced DA levels and decreased TH mRNA expression in mouse striata, effects that were significantly impeded by AAV2-hIL-10 administration prior to MPTP intoxication. AAV2-hIL-10 infusion increased IFN-γ, TGF-β and GFAP mRNA expression. Our data suggest that the transfer of AAV2-hIL-10 into the striatum may play a neuroprotective role in the mouse MPTP model of PD and these effects are mediated by the anti-inflammatory action of IL-10.
Product Description
Supelco
Taurine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Citric acid, Anhydrous, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Citric acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Taurine, ≥98%, FG
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
2-Hydroxybutyric acid sodium salt, 97%
Sigma-Aldrich
Citric acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Sodium phosphate, 96%
Citric acid, anhydrous, European Pharmacopoeia (EP) Reference Standard
Supelco
Citric acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
USP
Citric acid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Citric acid, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
Sigma-Aldrich
Citric acid, meets analytical specification of Ph. Eur., BP, USP, E330, anhydrous, 99.5-100.5% (based on anhydrous substance)
USP
Taurine, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.4-100.6%, powder
Sigma-Aldrich
Citric acid, anhydrous, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Citric acid, 99%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Taurine, suitable for cell culture, meets USP testing specifications
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Taurine, ≥99%
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
SAFC
Taurine
Sigma-Aldrich
Citric acid, BioUltra, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Taurine, BioUltra, ≥99.5% (T)