Merck
  • A simple and highly efficient method to identify the integration site of a transgene in the animal genome.
A simple and highly efficient method to identify the integration site of a transgene in the animal genome.
Neuroscience research (2014-01-01)
Shun Uemura, Tadahiro Nagaoka, Minesuke Yokoyama, Michihiro Igarashi, Masashi Kishi
ABSTRACT
Because genetic manipulation occasionally disrupts the expression of the neighboring genes, the chromosomal locus where the transgene has been integrated should be identified in the use of transgenic organisms. By using a new blend of thermostable DNA polymerase, we established a highly efficient method of inverse polymerase chain reaction for this purpose. By using this protocol, we successfully determined the vector integration sites of 2 mouse lines, NSE-tTA and tetO-Cre, the combination of which is a useful tool in neuroscience research. On the basis of this information, we quantified the relative expression amount of the chromosomal genes adjacent to these transgenes and found that the insertion of the tetO-Cre vector significantly altered the mRNA level of one of the examined genes. Considering the potential risk of the insertion effect, we recommend that the vector integration sites of any transgenic lines should be determined routinely by using this method, and that the expression levels of their neighboring genes should be determined.
Product Description
Supelco
Phenol solution, 5000 μg/mL in methanol, certified reference material
Supelco
Phenol solution, certified reference material, 500 μg/mL in methanol
Supelco
Phenol solution, 100 μg/mL in acetonitrile, PESTANAL®, analytical standard
Sigma-Aldrich
Phenol solution, Saturated with 0.01 M citrate buffer, pH 4.3 ± 0.2, BioReagent, Molecular Biology
Sigma-Aldrich
Liquified Phenol, ≥89.0%
Sigma-Aldrich
Phenol solution, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, BioReagent, Molecular Biology
Sigma-Aldrich
Phenol, ≥96.0% (calc. on dry substance, T)
Sigma-Aldrich
Phenol, Molecular Biology
Sigma-Aldrich
Phenol, BioXtra, ≥99.5% (GC)
USP
Phenol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Phenol, BioUltra, Molecular Biology, ≥99.5% (GC)
Supelco
Phenol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Phenol, natural, 97%, FG
Sigma-Aldrich
Phenol, ACS reagent, ≥99.0%
Sigma-Aldrich
Phenol, unstabilized, purified by redistillation, ≥99%
Sigma-Aldrich
Phenol, unstabilized, ReagentPlus®, ≥99%
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (GC)
Sigma-Aldrich
Phenol, ≥99%
Sigma-Aldrich
Phenol, puriss., ≥99.5% (GC), meets analytical specification of Ph. Eur., BP, USP, crystalline (detached)
Supelco
Phenol, PESTANAL®, analytical standard
Sigma-Aldrich
Phenol, BioUltra, Molecular Biology, TE-saturated, ~73% (T)
Sigma-Aldrich
Phenol, puriss. p.a., ACS reagent, reag. Ph. Eur., 99.0-100.5%