Merck
  • Increased dialysate MCP-1 is associated with cardiovascular mortality in peritoneal dialysis patients: a prospective observational study.
Increased dialysate MCP-1 is associated with cardiovascular mortality in peritoneal dialysis patients: a prospective observational study.
American journal of nephrology (2014-10-18)
Kwang Il Ko, Kyoung Sook Park, Mi Jung Lee, Fa Mee Doh, Chan Ho Kim, Hyang Mo Koo, Hyung Jung Oh, Jung Tak Park, Seung Hyeok Han, Shin-Wook Kang, Tae-Hyun Yoo
ABSTRACT
The aim of this study was to investigate the association between the dialysate MCP-1 (dMCP-1) and systemic inflammatory and nutritional markers in peritoneal dialysis (PD) patients. In addition, we examined the prognostic value of dMCP-1 on all-cause or cardiovascular mortality in these patients. We prospectively followed 169 prevalent PD patients from April 1st 2008 to December 31st 2012. At baseline, dMCP-1 and serum biochemical parameters including high sensitivity CRP (hs-CRP) and albumin were checked. All-cause mortality and cause of death were evaluated during the follow-up period. Based on the median level of dMCP-1, patients were classified as either low or high dMCP-1 groups. Mean age, hs-CRP, and D/Pcr ratio at 4 h were significantly higher, while serum albumin levels and %lean body mass (LBM) were significantly lower in the high dMCP-1 group. During the mean follow-up period of 47.7 months, all-cause mortality and cardiovascular mortality rate were significantly higher in the high dMCP-1 group (9.6 and 6.3 per 100 person-years, respectively) compared to the low dMCP-1 group (5.1 and 3.1 per 100 person-years, respectively; p = 0.021, 0.038). In multivariate Cox analysis, high dMCP-1 was a significant independent predictor of all-cause mortality (hazard ratio: 1.83, 95% confidence interval: 1.03-3.24, p = 0.039). dMCP-1 levels are closely correlated with nutritional and systemic inflammatory markers in PD patients. In addition, increased dMCP-1 is significantly associated with higher all-cause and cardiovascular mortality. These findings suggest that local peritoneal inflammation could contribute to poor clinical outcomes in PD patients.
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Urea, meets USP testing specifications
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Urea, BioUltra, Molecular Biology, 99% (T)
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Urea, suitable for electrophoresis
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Supelco
Urea, analytical standard
Sigma-Aldrich
Urea, powder, BioReagent, Molecular Biology, suitable for cell culture
USP
Urea, United States Pharmacopeia (USP) Reference Standard
Urea, European Pharmacopoeia (EP) Reference Standard
Millipore
Urea solution, suitable for microbiology, 40% in H2O
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O