Merck
  • De novo transcriptome sequencing in Pueraria lobata to identify putative genes involved in isoflavones biosynthesis.
De novo transcriptome sequencing in Pueraria lobata to identify putative genes involved in isoflavones biosynthesis.
Plant cell reports (2014-12-31)
Xin Wang, Shutao Li, Jia Li, Changfu Li, Yansheng Zhang
ABSTRACT
Using Illumina sequencing technology, we have generated the large-scale transcriptome sequencing data and indentified many putative genes involved in isoflavones biosynthesis in Pueraria lobata. Pueraria lobata, a member of the Leguminosae family, is a traditional Chinese herb which has been used since ancient times. P. lobata root has extensive clinical usages, because it contains a rich source of isoflavones, including daidzin and puerarin. However, the knowledge of isoflavone metabolism and the characterization of corresponding genes in such a pathway remain largely unknown. In this study, de novo transcriptome of P. lobata root and leaf was sequenced using the Solexa sequencing platform. Over 140 million high-quality reads were assembled into 163,625 unigenes, of which about 43.1% were aligned to the Nr protein database. Using the RPKM (reads per kilo bases per million reads) method, 3,148 unigenes were found to be upregulated, and 2,011 genes were downregulated in the leaf as compared to those in the root. Towards a further understanding of these differentially expressed genes, Gene ontology enrichment and metabolic pathway enrichment analyses were performed. Based on these results, 47 novel structural genes were identified in the biosynthesis of isoflavones. Also, 22 putative UDP glycosyltransferases and 45 O-methyltransferases unigenes were identified as the candidates most likely to be involved in the tailoring processes of isoflavonoid downstream pathway. Moreover, MYB transcription factors were analyzed, and 133 of them were found to have higher expression levels in the roots than in the leaves. In conclusion, the de novo transcriptome investigation of these unique transcripts provided an invaluable resource for the global discovery of functional genes related to isoflavones biosynthesis in P. lobata.
Sigma-Aldrich
3-Hydroxyflavone, ≥98%
Supelco
1,4-Benzoquinone, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Glycine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Glycine, analytical standard, for nitrogen determination according to Kjeldahl method
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Sigma-Aldrich
Glycine, 99%, FCC
SAFC
Glycine
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
USP
1,4-Benzoquinone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
p-Benzoquinone, reagent grade, ≥98%
Supelco
p-Benzoquinone, for spectrophotometric det. of amines, ≥99.5% (HPLC)
Supelco
Glycine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glycine, BioUltra, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Sigma-Aldrich
Glycine, puriss. p.a., reag. Ph. Eur., buffer substance, 99.7-101% (calc. to the dried substance)
Sigma-Aldrich
Glycine, tested according to Ph. Eur.
Sigma-Aldrich
Glycine, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
USP
Glycine, United States Pharmacopeia (USP) Reference Standard
Glycine, European Pharmacopoeia (EP) Reference Standard