Merck
  • Cognitive effects of vanillic acid against streptozotocin-induced neurodegeneration in mice.
Cognitive effects of vanillic acid against streptozotocin-induced neurodegeneration in mice.
Pharmaceutical biology (2014-12-05)
Jayasingh Chellammal Hanish Singh, Rahul Motiram Kakalij, Rahul Padmakar Kshirsagar, Boyina Hemanth Kumar, Sai Santosh Babu Komakula, Prakash Vamanrao Diwan
ABSTRACT
Vanillic acid (VA), a flavoring agent used in food and drug products, obtained naturally from the plant Angelica sinensis (Oliv.) Diels (Apiaceae), used in the traditional Chinese medicine. It is reported to possess strong antioxidant, anti-inflammatory, and neuroprotective effects. However, the pharmacological effects on oxidative stress-induced neurodegeneration are not well investigated. This study investigates the neuroprotective effect of VA on streptozotocin (STZ)-induced neurodegeneration in mice through behavioral and biochemical parameters. The behavioral effects were determined using the Y-maze and open-field habituation memory. In biochemical parameters, acetylcholinesterase (AChE), corticosterone, tumor necrosis factor (TNF)-α, and antioxidants (superoxide dismutase (SOD), glutathione peroxidase (GPx), and catalase) were measured. Five groups of animals used were of control, negative control, and three separate groups treated with 25, 50, and 100 mg/kg of VA, respectively, for 28 d. Intracerebroventricular (ICV) injections of STZ were performed for all groups except control on 14th and 16th of 28 d of VA treatment. VA improved spatial learning and memory retention by preventing oxidative stress compared with control animals. VA at 50 and 100 mg/kg dose significantly (p < 0.001) improved the habituation memory, decreased the AChE, corticosterone, TNF-α, and increased the antioxidants (p < 0.001). VA (100 mg/kg) exhibited dose-dependent effect in all parameters with p < 0.001 except antioxidants in which VA showed the significance of p < 0.01. VA exhibited reduction in AChE, TNF-α, and corticosterone with improved antioxidants to contribute neuroprotection and could be an effective therapeutic agent for treating neurodegenerative disorders.
Product Description
Dexamethasone, European Pharmacopoeia (EP) Reference Standard
Supelco
Adenosine, Pharmaceutical Secondary Standard; Certified Reference Material
Adenosine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Acetylthiocholine iodide, ≥98% (TLC), powder or crystals
USP
Dexamethasone, United States Pharmacopeia (USP) Reference Standard
Supelco
Dexamethasone, Pharmaceutical Secondary Standard; Certified Reference Material
Dexamethasone, British Pharmacopoeia (BP) Assay Standard
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%
Sigma-Aldrich
Adenosine, BioReagent, suitable for cell culture
Sigma-Aldrich
Acetylthiocholine iodide, ≥99.0% (AT)
Sigma-Aldrich
Adenosine, ≥99%
Sigma-Aldrich
Adenosine
Sigma-Aldrich
Magnesium chloride, ≥98%
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Magnesium chloride, suitable for insect cell culture, BioReagent, ≥97.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Dexamethasone, tested according to Ph. Eur.