Merck
  • Comparison of the Effectiveness of Nonablative Fractional Laser versus Pulsed-Dye Laser in Thyroidectomy Scar Prevention.
Comparison of the Effectiveness of Nonablative Fractional Laser versus Pulsed-Dye Laser in Thyroidectomy Scar Prevention.
Annals of dermatology (2014-10-18)
Ji Min Ha, Han Su Kim, Eun Byul Cho, Gyeong Hun Park, Eun Joo Park, Kwang Ho Kim, Lee Su Kim, Kwang Joong Kim
ABSTRACT
The anterior neck is the site of open thyroidectomy and where postoperative scarring can cause distress to patients. Both fractional and pulsed-dye lasers are effective and safe methods for preventing and improving surgical scars. This study evaluated the improvement in scar appearance with laser intervention during the wound healing process. We evaluated the effect of nonablative fractional and pulsed-dye lasers on fresh thyroidectomy scars. Patients were treated 3 times at 4-week interval with a follow-up visit at the 6(th) month. Scars were divided into 2 halves for each optional treatment. At every visit, a questionnaire evaluating the scar and patient satisfaction was completed. Thirty patients completed the 6-month process. The mean Vancouver Scar Scale scores improved significantly from 8.0 to 4.6 and 8.2 to 4.7 with nonablative fractional and pulsed-dye lasers, respectively (p<0.001). However, there was no significant difference between the 2 methods (p=0.840). There remains no consensus on the optimal treatment of scars. The present study indicates nonablative fractional and pulsed-dye lasers significantly improve scars. Nonablative fractional lasers are non-inferior to pulsed-dye lasers. Further studies are required to corroborate this finding.
Product Description
Lidocaine, European Pharmacopoeia (EP) Reference Standard
Supelco
Lidocaine, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Lidocaine, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Lidocaine, analytical standard
Sigma-Aldrich
Lidocaine, powder