Merck
  • Validated HILIC-MS/MS assay for determination of vindesine in human plasma: Application to a population pharmacokinetic study.
Validated HILIC-MS/MS assay for determination of vindesine in human plasma: Application to a population pharmacokinetic study.
Journal of pharmaceutical and biomedical analysis (2014-04-12)
Rong-Hua Zhu, Huan-De Li, Hua-Lin Cai, Zhi-Ping Jiang, Ping Xu, Li-Bo Dai, Wen-Xing Peng
ABSTRACT
The first HILIC-tandem mass spectrometry (MS/MS) method for determination of vindesine (VDS) in human plasma using vinorelbine as an internal standard (IS) has been developed and validated. Plasma samples clean-up consisted of solid phase extraction with a strata™-X column. The compounds were separated on a HILIC column with an isocratic mobile phase consisting of acetonitrile and 15mM ammonium acetate buffer containing 0.15% formic acid (80:20, v/v). The detection was performed on a triple quadrupole tandem mass spectrometer via electrospray positive ionization (ESI(+)). The ion transitions recorded in multiple reaction monitoring mode were m/z 754.6→123.8 for VDS and 779.4→323.3 for IS, respectively. Linear calibration curves were obtained in the concentration range of 0.3-28ng/ml and the lower limit of quantification for VDS was 0.3ng/ml. The coefficient of variation of the assay precision was less than 13%, and the accuracy exceeded 96%. The developed assay method was successfully applied for the evaluation of population pharmacokinetics of VDS after intravenous infusion of Xi Ai Ke Vial(®) (3mg of Vindesine Sulfate for Injection) to Chinese Han subjects with hematological malignant disorders.
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, Absolute - Acetone free
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Vindesine sulfate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile, ≥99.8% (GC)
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard